Skip to Content
Merck
All Photos(1)

Key Documents

SML2474

Sigma-Aldrich

HS56

≥98% (HPLC)

Synonym(s):

2-[[1-(3-Chlorophenyl)-4,5-dihydro-4-oxo-1H-pyrazolo[3,4-d]pyrimidin-6-yl]thio]acetonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€68.90
25 MG
€254.00

€68.90


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€68.90
25 MG
€254.00

About This Item

Empirical Formula (Hill Notation):
C13H8ClN5OS
CAS Number:
Molecular Weight:
317.75
UNSPSC Code:
12352200
NACRES:
NA.77

€68.90


Estimated to ship on26 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NC(SCC#N)=NC2=C1C=NN2C3=CC(Cl)=CC=C3

Biochem/physiol Actions

HS56 is a highly selective and potent dual Pim/DAPK3 inhibitor that significantly reduces contractile intensity and phosphorylation levels of myosin light chain (LC20) in rat caudal arterial smooth muscle strips. HS56 decreases blood pressure in spontaneously hypertensive mice without affecting heart rate.
highly selective and potent dual Pim/DAPK3 inhibitor that significantly reduces contractile intensity and phosphorylation levels of myosin light chain (LC20)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Carlson et al.
Cell chemical biology, 25(10), 1195-1207 (2018-07-24)
Sustained vascular smooth muscle hypercontractility promotes hypertension and cardiovascular disease. The etiology of hypercontractility is not completely understood. New therapeutic targets remain vitally important for drug discovery. Here we report that Pim kinases, in combination with DAPK3, regulate contractility and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service