Skip to Content
Merck
All Photos(1)

Key Documents

SML2468

Sigma-Aldrich

CINPA1

≥98%

Synonym(s):

CAR Inhibitor not PXR Activator 1, Ethyl 5-(2-(diethylamino)acetyl)-10,11-dihydro-5H-dibenzo[b,f]azepin-3-ylcarbamate, [5-[(Diethylamino)acetyl]-10,11-dihydro-5H-dibenz[b,f]azepin-3-yl]carbamic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€74.60
25 MG
€303.00

€74.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€74.60
25 MG
€303.00

About This Item

Empirical Formula (Hill Notation):
C23H29N3O3
CAS Number:
Molecular Weight:
395.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€74.60


Check Cart for Availability

Request a Bulk Order

Assay

≥98%

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CN(CC)CC)N1C2=C(C=CC=C2)CCC3=C1C=C(NC(OCC)=O)C=C3

InChI

1S/C23H29N3O3/c1-4-25(5-2)16-22(27)26-20-10-8-7-9-17(20)11-12-18-13-14-19(15-21(18)26)24-23(28)29-6-3/h7-10,13-15H,4-6,11-12,16H2,1-3H3,(H,24,28)

InChI key

AYQBYSPEGRYKFA-UHFFFAOYSA-N

Biochem/physiol Actions

CINPA1 is a ligand-binding domain (LBD)-targeting constitutive androstane receptor (CAR) inhibitor that prevents CAR recruitment to promoter regions of regulated genes (by >85%; 1 μM CINPA1 against 0.1 μM CITCO-induced CAR recruitment to CYP2B6 or CYP3A4 promoters; human hepatocytes) by altering CAR-coregulator interactions. CINPA1 inhibits CAR-mediated transcription in a potent (IC50 = 70 nM; HepG2-hCAR1 CYP2B6-luc reporter cells) and selective manner with only weak PXR antagonist potency (68% inhibition by 18 μM CINPA1 against 5 μM rifampicin; HepG2-PXR reporter cells) and no cytotoxicity or agonist/antagonist activity toward FXR, GR, LXRα/β, PPARγ, RXRα/β, VDR.
Potent and selective ligand-binding domain (LBD)-targeting constitutive androstane receptor (CAR) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenwei Lin et al.
European journal of medicinal chemistry, 108, 505-528 (2015-12-31)
Constitutive androstane receptor (CAR, NR1I3) and pregnane X receptor (PXR, NR1I2) are master regulators of endobiotic and xenobiotic metabolism and disposition. Because CAR is constitutively active in certain cellular contexts, inhibiting CAR might reduce drug-induced hepatotoxicity and resensitize drug-resistant cancer
Milu T Cherian et al.
Biochemical pharmacology, 152, 211-223 (2018-04-03)
The constitutive androstane receptor (CAR) and pregnane X receptor (PXR) are xenobiotic sensors that regulate the expression of drug-metabolizing enzymes and efflux transporters. CAR activation promotes drug elimination, thereby reducing therapeutic effectiveness, or causes adverse drug effects via toxic metabolites.
Baddreddine Boussadia et al.
Experimental neurology, 283(Pt A), 39-48 (2016-06-01)
Nuclear receptors (NRs) are a group of transcription factors emerging as players in normal and pathological CNS development. Clinically, an association between the constitutive androstane NR (CAR) and cognitive impairment was proposed, however never experimentally investigated. We wished to test
Milu T Cherian et al.
Drug metabolism and disposition: the biological fate of chemicals, 44(11), 1759-1770 (2016-08-16)
The constitutive androstane receptor (CAR) regulates the expression of genes involved in drug metabolism and other processes. A specific inhibitor of CAR is critical for modulating constitutive CAR activity. We recently described a specific small-molecule inhibitor of CAR, CINPA1 (ethyl
Judith Jeske et al.
Archives of toxicology, 91(6), 2375-2390 (2017-03-16)
Unintentional activation of xenosensing nuclear receptors pregnane X receptor (PXR) and/or constitutive androstane receptor (CAR) by clinical drug use is known to produce severe side effects in patients, which may be overcome by co-administering antagonists. However, especially antagonizing CAR is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service