Skip to Content
Merck
All Photos(1)

Key Documents

SML2424

Sigma-Aldrich

Terlipressin acetate salt

≥98% (HPLC)

Synonym(s):

H-Gly-Gly-Gly-Cys-Tyr-Phe-Gln-Asn-Cys-Pro-Lys-GlyNH2 acetate salt (Disulfide bond), N-(glycylglycylglycyl)-8-L-lysine-vasopressin acetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€87.30
25 MG
€355.00

€87.30


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€87.30
25 MG
€355.00

About This Item

Empirical Formula (Hill Notation):
C52H74N16O15S2 · xC2H4O2
CAS Number:
Molecular Weight:
1227.37 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€87.30


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

lyophilized powder

storage condition

desiccated

color

white

shipped in

wet ice

storage temp.

−20°C

SMILES string

S1SC[C@@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@@H](C1)C(=O)N4[C@@H](CCC4)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N)CC(=O)N)CCC(=O)N)Cc3ccccc3)Cc2ccc(cc2)O)NC(=O)CNC(=O)CNC(=O)CN.OC(=O)C.OC(=O)C

InChI

1S/C52H74N16O15S2.2C2H4O2/c53-17-5-4-9-31(45(76)60-23-41(57)72)63-51(82)38-10-6-18-68(38)52(83)37-27-85-84-26-36(61-44(75)25-59-43(74)24-58-42(73)22-54)50(81)65-34(20-29-11-13-30(69)14-12-29)48(79)64-33(19-28-7-2-1-3-8-28)47(78)62-32(15-16-39(55)70)46(77)66-35(21-40(56)71)49(80)67-37;2*1-2(3)4/h1-3,7-8,11-14,31-38,69H,4-6,9-10,15-27,53-54H2,(H2,55,70)(H2,56,71)(H2,57,72)(H,58,73)(H,59,74)(H,60,76)(H,61,75)(H,62,78)(H,63,82)(H,64,79)(H,65,81)(H,66,77)(H,67,80);2*1H3,(H,3,4)/t31-,32-,33-,34-,35-,36-,37-,38-;;/m0../s1

InChI key

WNFVFDPQEHRNTC-LWCZBKQBSA-N

Biochem/physiol Actions

Synthetic vasopressin analog and agonist
Terlipressin is a synthetic vasopressin analog and agonist. It has been used as a vasoactive drug for treatment of Hepatorenal syndrome in cirrhosis of the liver.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tim G Kampmeier et al.
Scientific reports, 8(1), 7105-7105 (2018-05-10)
The Surviving Sepsis Guidelines suggest the use of vasopressin in case of catecholamine-refractory septic shock. Terlipressin (TP) as a V1-selective AVP analogue is a potential alternative, though data regarding the first-line administration in septic shock are scarce. The present study
Timothy Papaluca et al.
Journal of gastroenterology and hepatology, 33(3), 591-598 (2017-10-06)
Terlipressin is an analogue of vasopressin that has potent vasoactive properties and has been available for use in most countries for nearly two decades. It has both established roles and emerging indications in the management of complications of decompensated chronic
K K Ida et al.
British journal of anaesthesia, 120(6), 1245-1254 (2018-05-26)
During early treatment of haemorrhagic shock, cerebral perfusion pressure can be restored by small-volume resuscitation with vasopressors. Whether this therapy is improved with additional fluid remains unknown. We assessed the value of terlipressin and lactated Ringer's solution (LR) on early

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service