Skip to Content
Merck
All Photos(1)

Key Documents

SML2406

Sigma-Aldrich

JI130

≥95% (HPLC)

Synonym(s):

(E)-N-(2-(Allyloxy)phenethyl)-3-(7-methoxy-1H-indol-3-yl)acrylamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€55.80
25 MG
€231.00

€55.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€55.80
25 MG
€231.00

About This Item

Empirical Formula (Hill Notation):
C23H24N2O3
CAS Number:
Molecular Weight:
376.45
UNSPSC Code:
12352200
NACRES:
NA.77

€55.80


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(/C=C/C1=CNC2=C1C=CC=C2OC)NCCC3=CC=CC=C3OCC=C

InChI

1S/C23H24N2O3/c1-3-15-28-20-9-5-4-7-17(20)13-14-24-22(26)12-11-18-16-25-23-19(18)8-6-10-21(23)27-2/h3-12,16,25H,1,13-15H2,2H3,(H,24,26)/b12-11+

InChI key

BNTYDBBGNPFECO-VAWYXSNFSA-N

Biochem/physiol Actions

JI130 is a cell penetrant, potent and selective inhibitor of the helix-loop-helix transcription factor Hes1 that blocks Hes1-mediated transcriptional repression. JI130 potently inhibits pancreatic cancer cell lines proliferation, and shrink tumor in a mice pancreatic tumor xenograft model.
cell penetrant, potent and selective inhibitor of the helix-loop-helix transcription factor Hes1 that blocks Hes1-mediated transcriptional repression

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelie Perron et al.
The Journal of biological chemistry, 293(21), 8285-8294 (2018-03-11)
The transcription factor Hes family basic helix-loop-helix transcription factor 1 (Hes1) is a downstream effector of Notch signaling and plays a crucial role in orchestrating developmental processes during the embryonic stage. However, its aberrant signaling in adulthood is linked to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service