Skip to Content
Merck
All Photos(1)

Key Documents

SML2385

Sigma-Aldrich

Andrastin A

≥95% (HPLC)

Synonym(s):

(3β,5β,8α,9β,10α,13α)-3β-(acetyloxy)-4,4,8,12,16-pentamethyl-15,17,19-trioxoandrost-11-ene-14-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€169.00

€169.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€169.00

About This Item

Empirical Formula (Hill Notation):
C28H38O7
CAS Number:
Molecular Weight:
486.60
MDL number:
UNSPSC Code:
12352200

€169.00


Estimated to ship on24 April 2025


Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

O(C1C(C2[C@@]([C@@H]3C(C4([C@](C(=C3)C)(C(=O)C(C4=O)C)C)C(=O)OC)(CC2)C)(CC1)C=O)(C)C)C(=O)C

InChI

1S/C28H38O7/c1-15-13-19-25(6,28(23(33)34-8)22(32)16(2)21(31)26(15,28)7)11-9-18-24(4,5)20(35-17(3)30)10-12-27(18,19)14-29/h13-14,16,18-20H,9-12H2,1-8H3/t16?,18?,19-,20?,25?,26+,27+,28?/m0/s1

InChI key

GRBXNADBNJGZRK-BCMOVVIDSA-N

Biochem/physiol Actions

Andrastin A is a meroterpenoid protein farnesyltransferase inhibitor with anticancer activity. It has been shown to inhibit the farnesyltransferase activity of the oncogenic Ras proteins and inhibit the efflux of anticancer drugs from multidrug-resistant cancer cells.
Farnesyltransferase inhibitor with anticancer activity; mycotoxin

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mytre Koul et al.
Anti-cancer drugs, 28(1), 11-30 (2016-08-24)
Secondary metabolites from fungal endophytes have become an interesting, attractive, and alternative source for novel pharmaceuticals. Several novel compounds with diversified chemical structures have been isolated from endophytic fungi. The genus Penicillium has been exploited worldwide for its biosynthetic potential
Juan F Rojas-Aedo et al.
Frontiers in microbiology, 8, 813-813 (2017-05-23)
Penicillium roqueforti is a filamentous fungus involved in the ripening of several kinds of blue cheeses. In addition, this fungus produces several secondary metabolites, including the meroterpenoid compound andrastin A, a promising antitumoral compound. However, to date the genomic cluster
Kristian Fog Nielsen et al.
Journal of agricultural and food chemistry, 53(8), 2908-2913 (2005-04-14)
This is the first finding of andrastins in blue cheese as well as any other sample type. Here, they were produced by the secondary starter culture Penicillium roqueforti. After purification by normal-phase chromatography followed by combined reverse-phase ion-exchange chromatography, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service