Skip to Content
Merck
All Photos(1)

Key Documents

SML2382

Sigma-Aldrich

SC75741

≥97% (HPLC)

Synonym(s):

4SC-301, N-(5-Benzoyl-1H-benzimidazol-2-yl)-2-(1-thieno[3,2-d]pyrimidin-4-yl-4-piperidinyl)-4-thiazolecarboxamide, N-(6-Benzoyl-1H-benzimidazol-2-yl)-2-(1-thieno[3,2-d]pyrimidin-4-yl-4-piperidinyl)-4-thiazolecarboxamide, SC 75741, V1810

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€65.60
25 MG
€265.00

€65.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€65.60
25 MG
€265.00

About This Item

Empirical Formula (Hill Notation):
C29H23N7O2S2
CAS Number:
Molecular Weight:
565.67
UNSPSC Code:
12352200

€65.60


Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CSC(C2CCN(C3=C4C(C=CS4)=NC=N3)CC2)=N1)NC5=NC6=CC(C(C7=CC=CC=C7)=O)=CC=C6N5

InChI

1S/C29H23N7O2S2/c37-24(17-4-2-1-3-5-17)19-6-7-20-22(14-19)34-29(33-20)35-27(38)23-15-40-28(32-23)18-8-11-36(12-9-18)26-25-21(10-13-39-25)30-16-31-26/h1-7,10,13-16,18H,8-9,11-12H2,(H2,33,34,35,38)

InChI key

QNZVBFMXWNWVKG-UHFFFAOYSA-N

Biochem/physiol Actions

NF-κB p65 subunit (RelA) DNA-binding inhibitor that effectively blocks influenza virus propagation both in cultures and in mice in vivo.
SC75741 (4SC-301; V1810) is a potent NF-kappaB (NF-κB) inhibitor (IC50 = 200 nM against 10 ng/mL TNF-α-induced NF-κB reporter gene expression in A549 cells) that impairs p65 subunit (RelA) DNA-binding activity without affecting p65 phosphorylation or nuclear translocation, nor proteasome-mediated IκBα degradation. SC75741 induces apoptosis in multiple myeloma cultures (IC50 from 5 to 12 μM in 48 hrs; OPM2, U266, NCI-H929, RPMI-8226), but not PBMCs (89% viability post 4-hr 100 μM SC75741 treatment). NF-κB inhibition by SC75741 treatment is reported to effectively block influenza virus propagation both in cultures in vitro (1-5 μM) and in mice in vivo (5 mg/kg daily i.v. or 7.5-15 mg/kg twice a day i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenshuang Wu et al.
Oncotarget, 7(32), 52281-52293 (2016-07-23)
Millepachine (MIL) was a novel chalcone that was separated from Millettia pachycarpa Benth (Leguminosae). We found MIL induced apoptosis through activating NF-κB pathway both in SK-OV-3 and A2780S cells. Western blot showed that MIL increased the levels of IKKα, p-IKKα/β
Ke-Wang Luo et al.
The Journal of nutritional biochemistry, 41, 56-64 (2017-01-04)
Epigallocatechin-3-gallate (EGCG), the bioactive polyphenol in green tea, has been demonstrated to have various biological activities. Our study aims to investigate the antiproliferation and antimigration effects of EGCG against bladder cancer SW780 cells both in vitro and in vivo. Our
Christina Ehrhardt et al.
Cellular microbiology, 15(7), 1198-1211 (2013-01-17)
Ongoing human infections with highly pathogenic avian H5N1 viruses and the emergence of the pandemic swine-origin influenza viruses (IV) highlight the permanent threat elicited by these pathogens. Occurrence of resistant seasonal and pandemic strains against the currently licensed antiviral medications
Johann Leban et al.
Bioorganic & medicinal chemistry letters, 17(21), 5858-5862 (2007-09-18)
A novel class of NF-kappaB pathway signaling inhibitors was discovered by virtual screening, medicinal chemistry, and QSAR analysis. An initial set of compounds inhibited NF-kappaB signaling in a whole cell reporter gene assay in the micro-molar range. Activity was improved
Emanuel Haasbach et al.
Antiviral research, 99(3), 336-344 (2013-07-03)
The appearance of pandemic H1N1 and highly pathogenic avian H5N1 viruses in humans as well as the emergence of seasonal H1N1 variants resistant against neuraminidase inhibitors highlight the urgent need for new and amply available antiviral drugs. We and others

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service