Skip to Content
Merck
All Photos(1)

Key Documents

SML2375

Sigma-Aldrich

KDU691

≥98% (HPLC)

Synonym(s):

KDU 691, N-(4-Chlorophenyl)-N-methyl-3-[4-(methylcarbamoyl)phenyl]imidazo[1,2-a]pyrazine-6-carboxamide, N-(4-Chlorophenyl)-N-methyl-3-[4-[(methylamino)carbonyl]phenyl]imidazo[1,2-a]pyrazine-6-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€422.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€422.00

About This Item

Empirical Formula (Hill Notation):
C22H18ClN5O2
CAS Number:
Molecular Weight:
419.86
MDL number:
UNSPSC Code:
12352200

€112.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N(C1=CC=C(Cl)C=C1)C)C(N=C2)=CN3C2=NC=C3C4=CC=C(C(NC)=O)C=C4

InChI

1S/C22H18ClN5O2/c1-24-21(29)15-5-3-14(4-6-15)19-11-26-20-12-25-18(13-28(19)20)22(30)27(2)17-9-7-16(23)8-10-17/h3-13H,1-2H3,(H,24,29)

InChI key

TYMFFISSODJRDV-UHFFFAOYSA-N

Biochem/physiol Actions

KDU691 is an orally active imidazopyrazine class antiparasitic that inhibits Plasmodium & Cryptosporidium phosphatidylinositol-4-OH kinase, PI(4)K, in an ATP-competitive, highly potent and selective manner (IC50/[ATP] = 1.5 nM/10 μM/P. vivax & 17 nM/3 μM/C. parvum PI(4)K) with little or no activity against human PI3Kα/β/γ/δ, PI4KIIIβ, VPS34, and 36 human protein kinases. KDU691 is effective against human pathogens P. falciparum, P. vivax, C. parvum and C. hominis, as well as simian parasite P. cynomolgi. KDU691 blocks Plasmodium development in all life-cycle stages and displays in vivo efficacy in murine models of malaria and cryptosporidiosis.
Orally active antiparasitic (antimalarial & anti-cryptosporidial), Plasmodium & Cryptosporidium phosphatidylinositol-4-OH kinase, PI(4)K, inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Case W McNamara et al.
Nature, 504(7479), 248-253 (2013-11-29)
Achieving the goal of malaria elimination will depend on targeting Plasmodium pathways essential across all life stages. Here we identify a lipid kinase, phosphatidylinositol-4-OH kinase (PI(4)K), as the target of imidazopyrazines, a new antimalarial compound class that inhibits the intracellular
L Dembele et al.
Scientific reports, 7(1), 2325-2325 (2017-05-26)
Malaria control and elimination are threatened by the emergence and spread of resistance to artemisinin-based combination therapies (ACTs). Experimental evidence suggests that when an artemisinin (ART)-sensitive (K13 wild-type) Plasmodium falciparum strain is exposed to ART derivatives such as dihydroartemisinin (DHA)
Laurent Dembele et al.
Antimicrobial agents and chemotherapy, 62(5) (2018-03-14)
Artemisinin (ART) resistance has spread through Southeast Asia, posing a serious threat to the control and elimination of malaria. ART resistance has been associated with mutations in the Plasmodium falciparum kelch-13 (Pfk13) propeller domain. Phenotypically, ART resistance is defined as
Anne-Marie Zeeman et al.
Antimicrobial agents and chemotherapy, 60(5), 2858-2863 (2016-03-02)
Two Plasmodium PI4 kinase (PI4K) inhibitors, KDU691 and LMV599, were selected for in vivo testing as causal prophylactic and radical-cure agents for Plasmodium cynomolgi sporozoite-infected rhesus macaques, based on their in vitro activity against liver stages. Animals were infected with
Koen J Dechering et al.
Scientific reports, 7(1), 17680-17680 (2017-12-17)
Eradication of malaria requires a novel type of drug that blocks transmission from the human to the mosquito host, but selection of such a drug is hampered by a lack of translational models. Experimental mosquito infections yield infection intensities that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service