Skip to Content
Merck
All Photos(1)

Key Documents

SML2360

Sigma-Aldrich

LX-4

≥98% (HPLC)

Synonym(s):

10(9H)-Acridineacetic acid, 9-oxo-, [[4-(2-benzothiazolylthio)-3-nitrophenyl]methylene]hydrazide, 9-Oxo-10(9H)-acridineacetic acid 2-[[4-(2-benzothiazolylthio)-3-nitrophenyl]methylene]hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€80.60
25 MG
€260.00

€80.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€80.60
25 MG
€260.00

About This Item

Empirical Formula (Hill Notation):
C29H19N5O4S2
CAS Number:
Molecular Weight:
565.62
UNSPSC Code:
12352200
NACRES:
NA.77

€80.60


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(nc1Sc3c(cc(cc3)\C=N\NC(=O)CN4c5c(cccc5)C(=O)c6c4cccc6)[N+](=O)[O-])cccc2

InChI

1S/C29H19N5O4S2/c35-27(17-33-22-10-4-1-7-19(22)28(36)20-8-2-5-11-23(20)33)32-30-16-18-13-14-26(24(15-18)34(37)38)40-29-31-21-9-3-6-12-25(21)39-29/h1-16H,17H2,(H,32,35)/b30-16+

InChI key

HUHDFULZZUYTIU-OKCVXOCRSA-N

Biochem/physiol Actions

LX-4 is a cell penetrant, potent and selective activator of p38 mitogen-activated protein kinase (MAPK) pathway that derepress a subset of endogenous genes repressed by DNA methylation.
cell penetrant, potent and selective activator of p38 mitogen-activated protein kinase (MAPK) pathway

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service