Skip to Content
Merck
All Photos(1)

Key Documents

SML2301

Sigma-Aldrich

EpoY

≥97% (HPLC)

Synonym(s):

(2S,3S)-3-[[[(1S)-1-Carboxy-2-(4-hydroxyphenyl)ethyl]amino]carbonyl]-2-oxiranecarboxylic acid 2-ethyl ester, (S)-2-((2S,3S)-3-(ethoxycarbonyl) oxirane -2-carboxamido)-3-(4-hydroxyphenyl) propanoic acid, SD-142

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€82.20
25 MG
€331.00

€82.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€82.20
25 MG
€331.00

About This Item

Empirical Formula (Hill Notation):
C15H17NO7
CAS Number:
Molecular Weight:
323.30
UNSPSC Code:
12352200

€82.20


Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@@H](CC1=CC=C(O)C=C1)C(O)=O)[C@H]2O[C@@H]2C(OCC)=O

InChI

1S/C15H17NO7/c1-2-22-15(21)12-11(23-12)13(18)16-10(14(19)20)7-8-3-5-9(17)6-4-8/h3-6,10-12,17H,2,7H2,1H3,(H,16,18)(H,19,20)/t10-,11-,12-/m0/s1

InChI key

PSLWNPKMALERSK-SRVKXCTJSA-N

Biochem/physiol Actions

EpoY is an irreversible inhibitor of the major brain tubulin tyrosine carboxypeptidase (TCP), shown to be a complex of vasohibin-1 (VASH1) with the small vasohibin binding protein (SVBP). Reversible detyrosination of alpha-tubulin is vital to microtubule dynamics and functions including regulation of neuron differentiation, and defects in the detryosination have also been been associated with cancer and cardiomyopathies EpoY inhibited TCP activity with an IC50 value around 500 nM. It reduced detyrosinated a-tubulin levels and caused severe differentiation defects.
Inhibitor of Vasohibin/SVBP, a tubulin tyrosine carboxypeptidase (TCP) that regulates neuron differentiation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chrystelle Aillaud et al.
Science (New York, N.Y.), 358(6369), 1448-1453 (2017-11-18)
Reversible detyrosination of α-tubulin is crucial to microtubule dynamics and functions, and defects have been implicated in cancer, brain disorganization, and cardiomyopathies. The identity of the tubulin tyrosine carboxypeptidase (TCP) responsible for detyrosination has remained unclear. We used chemical proteomics

Articles

Discover definitions and uses for irreversible inhibitors including the types of irreversible inhibitors: suicide inhibitors, heavy metal inhibitors, and time-dependent inhibitors.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service