Skip to Content
Merck

Skip To

SML2299

Photoregulin3

≥98% (HPLC)

Synonym(s):

2-[(3,6-Dihydro-4-phenyl-1(2H)-pyridinyl)methyl]-6,7-dimethoxy-4(3H)-quinazolinone, PR3

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H23N3O3
CAS Number:
Molecular Weight:
377.44
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(C1=C2)=NC(CN3CCC(C4=CC=CC=C4)=CC3)=NC1=CC(OC)=C2OC

InChI

1S/C22H23N3O3/c1-27-19-12-17-18(13-20(19)28-2)23-21(24-22(17)26)14-25-10-8-16(9-11-25)15-6-4-3-5-7-15/h3-8,12-13H,9-11,14H2,1-2H3,(H,23,24,26)

InChI key

CKCSOMMBIPJYSG-UHFFFAOYSA-N

Biochem/physiol Actions

Nr2e3 modulator
Photoregulin3 is a modulator of the rod transcription factor Nr2e3 that inhibits rod photoreceptor gene expression. Photoregulin3 significantly rescues photoreceptor in RhoP23H mice.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8736SML1145SML1085
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: ≥20 mg/mL

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear (warmed)

color

white to beige

color

yellow

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters