Skip to Content
Merck
All Photos(1)

Key Documents

SML2296

Sigma-Aldrich

GSK180736A

≥98% (HPLC)

Synonym(s):

4-(4-Fluorophenyl)-1,2,3,4-tetrahydro-N-1H-indazol-5-yl-6-methyl-2-oxo-5-pyrimidinecarboxamide, GSK 180736A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€74.20
25 MG
€241.00

€74.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€74.20
25 MG
€241.00

About This Item

Empirical Formula (Hill Notation):
C19H16FN5O2
CAS Number:
Molecular Weight:
365.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€74.20


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(NN=C2)C2=C1)C3=C(C)NC(NC3C4=CC=C(F)C=C4)=O

InChI

1S/C19H16FN5O2/c1-10-16(17(24-19(27)22-10)11-2-4-13(20)5-3-11)18(26)23-14-6-7-15-12(8-14)9-21-25-15/h2-9,17H,1H3,(H,21,25)(H,23,26)(H2,22,24,27)

InChI key

HEAIGWIZTYAQTC-UHFFFAOYSA-N

Biochem/physiol Actions

GSK180736A is a potent and selective dual inhibitor of a ROCK1 (Rho-associated, coiled-coil-containing protein kinase) and GRK2 (G protein-coupled receptor kinase 2). GSK180736A binds to the GRK2 active site. GSK180736A produces a significant cardiomyocytes contraction at 1 μM.
potent and selective dual inhibitor of a ROCK1 and GRK2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristoff T Homan et al.
ACS chemical biology, 10(1), 310-319 (2014-09-23)
Selective inhibitors of individual subfamilies of G protein-coupled receptor kinases (GRKs) would serve as useful chemical probes as well as leads for therapeutic applications ranging from heart failure to Parkinson's disease. To identify such inhibitors, differential scanning fluorimetry was used
Clark A Sehon et al.
Journal of medicinal chemistry, 51(21), 6631-6634 (2008-10-10)
Recent studies using known Rho-associated kinase isoform 1 (ROCK1) inhibitors along with cellular and molecular biology data have revealed a pivotal role of this enzyme in many aspects of cardiovascular function. Here we report a series of ROCK1 inhibitors which
Renee Bouley et al.
Molecular pharmacology, 92(6), 707-717 (2017-10-27)
G protein-coupled receptor kinases (GRKs) phosphorylate activated receptors to promote arrestin binding, decoupling from heterotrimeric G proteins, and internalization. GRK2 and GRK5 are overexpressed in the failing heart and thus have become therapeutic targets. Previously, we discovered two classes of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service