Skip to Content
Merck

Skip To

SML2295

Dimethyoxy-etomidate

≥98% (HPLC)

Synonym(s):

(R)-ethyl 1-(1-(3,5-dimethoxyphenyl)ethyl)-1H-imidazole-5-carboxylate

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H20N2O4
CAS Number:
Molecular Weight:
304.34
UNSPSC Code:
12352200
NACRES:
NA.21
Assay:
≥98% (HPLC)
Form:
oil


assay

≥98% (HPLC)

form

oil

color

colorless to light brown

storage temp.

2-8°C

SMILES string

C[C@@H](N1C=NC=C1C(OCC)=O)C2=CC(OC)=CC(OC)=C2

InChI

1S/C16H20N2O4/c1-5-22-16(19)15-9-17-10-18(15)11(2)12-6-13(20-3)8-14(7-12)21-4/h6-11H,5H2,1-4H3/t11-/m1/s1

InChI key

JETTTZITBCWAQX-LLVKDONJSA-N

Biochem/physiol Actions

Dimethyoxy-etomidate is a potent suppressor of adrenocortical steroid synthesis by inhibiting steroid 11β-hydroxylase. Dimethyoxy-etomidate is essentially divided of etomidate′s GABAA receptor positive modulatory and sedative-hypnotic activities.
potent suppressor of adrenocortical steroid synthesis, selective inhibitor of steroid 11β-hydroxylase
Dimethyoxy-etomidate is a phenyl ring substituted etomidate analog.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters