Skip to Content
Merck

Skip To

SML2293

NSC61610

≥98% (HPLC)

Synonym(s):

CID247228, N1,N4-Bis[3-(1H-benzimidazol-2-yl)phenyl]-1,4-benzenedicarboxamide, NCI 61610

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 251,00

About This Item

Empirical Formula (Hill Notation):
C34H24N6O2
CAS Number:
Molecular Weight:
548.59
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 251,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

off-white to purple

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C=C1)C(NC2=CC=CC(C3=NC4=C(N3)C=CC=C4)=C2)=O)NC5=CC(C6=NC7=C(N6)C=CC=C7)=CC=C5

InChI

1S/C34H24N6O2/c41-33(35-25-9-5-7-23(19-25)31-37-27-11-1-2-12-28(27)38-31)21-15-17-22(18-16-21)34(42)36-26-10-6-8-24(20-26)32-39-29-13-3-4-14-30(29)40-32/h1-20H,(H,35,41)(H,36,42)(H,37,38)(H,39,40)

InChI key

WSXIJTIFOWCMIQ-UHFFFAOYSA-N

Biochem/physiol Actions

NSC61610 is an orally available and potent lanthionine synthetase C-like 2 (LANCL2) ligand that induces IL-10-mediated anti-inflammatory responses that ameliorate disease activity and decrease inflammatory lesions in the gut. Also NSC61610 modulates immune response to influenza A H1N1pdm virus infection in mice.
orally available and potent lanthionine synthetase C-like 2 (LANCL2) ligand

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0634SML2445SML2631
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

off-white to purple

color

white to beige

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters