Skip to Content
Merck

Skip To

SML2235

CL075

≥98% (HPLC)

Synonym(s):

2-Propylthiazolo[4,5-c]quinolin-4-amine, 3M002

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 98,40
25 mg
Check Cart for Availability
€ 444,00

About This Item

Empirical Formula (Hill Notation):
C13H13N3S
CAS Number:
Molecular Weight:
243.33
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder

€ 98,40


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c2c(nc1CCC)c(nc3c2cccc3)N

InChI

1S/C13H13N3S/c1-2-5-10-16-11-12(17-10)8-6-3-4-7-9(8)15-13(11)14/h3-4,6-7H,2,5H2,1H3,(H2,14,15)

InChI key

NFYMGJSUKCDVJR-UHFFFAOYSA-N

Application

CL075 has been used to activate the monocyte-derived dendritic cells (DCs) to demonstrate the ability of DCs to induce cell death.

Biochem/physiol Actions

CL075 is a selective agonist of Toll-like receptor-8 (TLR-8) with immunostimulant activity. It showed an EC50 value of 400nM for IRF-NFΚB stimulation in TLR8 compared to 4 μM for TLR7.
CL075 is a thiazoloquinolone derivative[1]. It might influence tumor development.[2]
Toll-like receptor-8 (TLR-8) agonist

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2566SML0376SML2495
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (warmed)

solubility

DMSO: >2 mg/mL (warmed)

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to very dark brown

color

light yellow to light brown

color

white to gray-brown


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters