Skip to Content
Merck

Skip To

SML2226

BC-LI-0186

≥98% (HPLC)

Synonym(s):

4-(4-Isopropyl-2,3-dimethyl-5-oxo-2,5-dihydro-1H-pyrazol-1-yl)-N-(2-phenoxyethyl)benzenesulfonamide, 4-[2,5-Dihydro-2,3-dimethyl-4-(1-methylethyl)-5-oxo-1H-pyrazol-1-yl]-N-(2-phenoxyethyl)benzenesulfonamide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H27N3O4S
CAS Number:
Molecular Weight:
429.53
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(C(C)C)=C(C)N1C)N1C2=CC=C(S(NCCOC3=CC=CC=C3)(=O)=O)C=C2

InChI

1S/C22H27N3O4S/c1-16(2)21-17(3)24(4)25(22(21)26)18-10-12-20(13-11-18)30(27,28)23-14-15-29-19-8-6-5-7-9-19/h5-13,16,23H,14-15H2,1-4H3

InChI key

SQYWMHPZMHDCHP-UHFFFAOYSA-N

Biochem/physiol Actions

BC-LI-0186 is a selective blocker of Leucyl-tRNA synthetase (LRS; LeuRS) and Ras-related GTP-binding protein D (RagD) interaction (IC50 = 46.11 nM) by competing against RagD for LRS VC domain binding (KD = 42.1 nM) without affecting LRS-Vps34, LRS-EPRS, RagB-RagD association, mTORC1 complex formation or the activities of 12 kinases. BC-LI-0186 inhibits Leu-dependent LRS lysosomal membrane localization (10 μM), RagD GTPase and mTORC1 activation (IC50 = 81.4 nM against Leu-induced S6K phosphorylation), but not ARF1 activation, AKT S473 phosphorylation, Glu- or Arg-dependent S6K phosphorylation, nor the cytosolic and mitochondrial LRS catalytic activities. BC-LI-0186 effectively suppresses the growth of rapamycin-resistant MCT116 MM cancer cells with mTOR-S2035I mutation both in cultures (GI50 = 42.03) and in mice in vivo (by 40% in 2 wks; 20 mg/kg/day i.p.).
Leucyl-tRNA synthetase (LRS) and RagD interaction blocker that inhibits Leu-dependent LRS lysosomal localization, RagD and mTOR activation.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters