Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 mg | Check Cart for Availability | € 333,00 € 283,05 |
About This Item
Empirical Formula (Hill Notation):
C22H29N3O3S3
CAS Number:
Molecular Weight:
479.68
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
oil
Storage condition:
desiccated
€ 283,05
List Price€ 333,00Save 15%Web-Only Promotion
Check Cart for Availability
assay
≥98% (HPLC)
form
oil
storage condition
desiccated
color
colorless to light brown
storage temp.
2-8°C
SMILES string
O=C(CCCC[C@@H]1CCSS1)N2CCN(S(C3=CC=CC4=C3C=CN=C4)(=O)=O)CCC2
InChI
1S/C22H29N3O3S3/c26-22(8-2-1-6-19-10-16-29-30-19)24-12-4-13-25(15-14-24)31(27,28)21-7-3-5-18-17-23-11-9-20(18)21/h3,5,7,9,11,17,19H,1-2,4,6,8,10,12-16H2/t19-/m1/s1
InChI key
NWTRUKNNHUXONB-LJQANCHMSA-N
Biochem/physiol Actions
L-F001 is a multifunctional, brain penetrant and potent Rho-associated protein kinase (ROCK) inhibitor that protects the PC12 cells from paraquat induced cytotoxicity. L-F001 suppresses neuroinflammation in vitro and in vivo.
multifunctional, brain penetrant and potent Rho-associated protein kinase (ROCK) inhibitor
1 of 1
This Item | |||
|---|---|---|---|
| description ≥98% (HPLC) | description ≥98% (HPLC), solid | description powder | description ≥98% (HPLC) |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (TLC) | assay ≥98% (HPLC) |
| form oil | form solid | form powder | form powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| storage condition desiccated | storage condition - | storage condition - | storage condition desiccated |
| color colorless to light brown | color white | color white | color off-white to brown |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



