Skip to Content
Merck

Skip To

SML2153

TAM1 hydrochloride

≥98% (HPLC)

Synonym(s):

5-Hydroxy-4-[(4-methyl-1-piperidinyl)methyl]-2-phenyl-3-benzofurancarboxylic acid ethyl ester Hydrochloride

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C24H27NO4 · HCl
CAS Number:
Molecular Weight:
429.94
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

TAM1 is an inhibitor of the thioesterase activity of Mycobacterium tuberculosis polyketide synthase Pks13. TAM1 binds to Pks13 TE domain. TAM1 exhibit a potent bactericidal activity against Mycobacterium Tuberculosis H37Rv strain.
inhibitor of the thioesterase activity of Mycobacterium tuberculosis polyketide synthase Pks13.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters