Skip to Content
Merck
All Photos(1)

Key Documents

SML2065

Sigma-Aldrich

Mequitazine

≥98% (HPLC)

Synonym(s):

10-(1-Azabicyclo[2.2.2]oct-3-ylmethyl)-10H-phenothiazine, 10-(3-quinuclidinylmethyl) phenothiazine, LM 209, NSC 303612

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€86.80
25 MG
€297.00

€86.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€86.80
25 MG
€297.00

About This Item

Empirical Formula (Hill Notation):
C20H22N2S
CAS Number:
Molecular Weight:
322.47
UNSPSC Code:
12352200
NACRES:
NA.77

€86.80


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

S1c2c(cccc2)N(c5c1cccc5)CC3C4CCN(C3)CC4

InChI

1S/C20H22N2S/c1-3-7-19-17(5-1)22(18-6-2-4-8-20(18)23-19)14-16-13-21-11-9-15(16)10-12-21/h1-8,15-16H,9-14H2

InChI key

HOKDBMAJZXIPGC-UHFFFAOYSA-N

Biochem/physiol Actions

Mequitazine is a potent, nonsedative, long-acting and selective H1 histamine receptor antagonist. Mequitazine exhibits weak anticholinergic activity.
potent, nonsedative, long-acting and selective H1 histamine receptor antagonist

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Martinez-Mir et al.
The Journal of pharmacy and pharmacology, 40(9), 655-656 (1988-09-01)
The antihistamine and anticholinergic properties of mequitazine have been investigated and compared with those of clemizole. Both mequitazine and clemizole antagonized the effect of histamine in guinea-pig ileum competitively, the pA2 values calculated by Schild plot were 9.95 +/- 0.44
Tatsuya Isomura et al.
PloS one, 9(12), e114336-e114336 (2014-12-17)
Second-generation antihistamines (AHs) have, in general, fewer sedative effects than the first-generation. However, important inter-drug differences remain in the degree of cognitive and/or psychomotor impairment. The extent to which a particular compound causes disruption can be conveniently compared, to all
K Nakamura et al.
The Journal of pharmacology and experimental therapeutics, 284(2), 437-442 (1998-03-07)
Mequitazine [10-(3-quinuclidinylmethyl) phenothiazine] is a long-acting and selective histamine H1-receptor antagonist that is mainly biotransformed by human liver microsomes to yield hydroxylated and S-oxidized metabolites. Mequitazine hydroxylase was inhibited by propranolol and quinidine. Lineweaver-Burk plots for the hydroxylation and the
Shigeru Hishinuma et al.
Journal of pharmacological sciences, 107(1), 66-79 (2008-05-01)
We evaluated changes in the binding properties of sedative and non-sedative histamine H1-receptor antagonists induced by internalization of H1 receptors in intact human U373 MG astrocytoma cells. Internalization of H1 receptors was induced without their degradation by treatment with 0.1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service