Skip to Content
Merck
All Photos(1)

Key Documents

SML2052

Sigma-Aldrich

LCAT activator compound A

≥98% (HPLC)

Synonym(s):

3-[[5-(Ethylthio)-1,3,4-thiadiazol-2-yl]thio]-2-pyrazinecarbonitrile, 3-[[5-(Ethylthio)-1,3,4-thiadiazol-2-yl]thio]pyrazine-2-carbonitrile, Compound A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€71.50
25 MG
€291.00

€71.50


Check Cart for Availability


Select a Size

Change View
5 MG
€71.50
25 MG
€291.00

About This Item

Empirical Formula (Hill Notation):
C9H7N5S3
CAS Number:
Molecular Weight:
281.38
UNSPSC Code:
41106609
NACRES:
NA.77

€71.50


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CCSC1=NN=C(S1)SC2=NC=CN=C2C#N

InChI

1S/C9H7N5S3/c1-2-15-8-13-14-9(17-8)16-7-6(5-10)11-3-4-12-7/h3-4H,2H2,1H3

InChI key

UBSFSJMIJHSGDZ-UHFFFAOYSA-N

Biochem/physiol Actions

LCAT activator compound A is an activator of Lecithin:cholesterol acyltransferase (LCAT) including some mutant LCATs found in Familial LCAT Deficiency (FLD). LCAT activator compound A forms a covalent hydrophobic adduct with LCAT Cys 31. The chronic treatment with LCAT activator compound A results in a significant increase in HDLc, HDL particle, size, plasma apolipoprotein A-I level, plasma cholesteryl ester to free cholesterol ratio, and a significant reduction in very low-density lipoprotein cholesterol in hamsters.
Lecithin:cholesterol acyltransferase (LCAT) including some FLD mutant LCATs

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhu Chen et al.
Metabolism: clinical and experimental, 61(4), 470-481 (2011-10-18)
The objective was to assess whether pharmacological activation of lecithin cholesterol acyltransferase (LCAT) could exert beneficial effects on lipoprotein metabolism. A putative small molecule activator (compound A) was used as a tool compound in in vitro and in vivo studies.
Lita A Freeman et al.
The Journal of pharmacology and experimental therapeutics, 362(2), 306-318 (2017-06-04)
Lecithin:cholesterol acyltransferase (LCAT) catalyzes plasma cholesteryl ester formation and is defective in familial lecithin:cholesterol acyltransferase deficiency (FLD), an autosomal recessive disorder characterized by low high-density lipoprotein, anemia, and renal disease. This study aimed to investigate the mechanism by which compound

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service