Skip to Content
Merck
All Photos(1)

Key Documents

SML1999

Sigma-Aldrich

Sm4

≥98% (HPLC)

Synonym(s):

2-Hydroxy-6-(2-(naphthalen-2-yl)ethyl)benzoic acid, 2-Hydroxy-6-[2-(2-naphthyl)ethyl]benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€127.00
25 MG
€512.00

€127.00


Check Cart for Availability


Select a Size

Change View
5 MG
€127.00
25 MG
€512.00

About This Item

Empirical Formula (Hill Notation):
C19H16O3
CAS Number:
Molecular Weight:
292.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€127.00


Check Cart for Availability

Assay

≥98% (HPLC)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C(O)=O)C(CCC2=CC3=C(C=CC=C3)C=C2)=CC=C1

InChI

1S/C19H16O3/c20-17-7-3-6-15(18(17)19(21)22)11-9-13-8-10-14-4-1-2-5-16(14)12-13/h1-8,10,12,20H,9,11H2,(H,21,22)

InChI key

GGVFBUYNEPBFQT-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active SOX18 inhibitor with in vivo efficacy against zebrafish larvae vascular formation and breast cancer tumor lymphangiogenesis in mice.
Sm4 is an orally active SOX18 inhibitor (IC50 = 5,2 μM; COS-7 reporter assay) that directly targets SOX18 HMG domain and preferentially disrupts SOX18 interaction with a subset of binding partners (IC50 = 3.3 μM/SOX18-SOX18, 15.8 μM/SOX18-DDX17, 42.3 μM/SOX18-RBPJ, 65.9 μM/SOX18-RBPJ; IC50 ≥780 μM against SOX9 dimer or SOX18 interaction with MEF2C, NR2F2, TRIM28, ESR1), while affecting HMG-dependent DNA binding in a less potent and non-SOX18-selective manner (IC50 ∼200-220 μM for SOX15/18, IC50 ∼270-310 μM for SOX2/6/9/11). Sm4 selectively affects SOX18 target genes transcription over 7 other transcription factors by genome-wide ChIP-seq analysis as well as displays in vivo efficacy against vascular formation in zebrafish larvae (1-2 μM) and induces tumor metastasis by inhibiting angiogenesis/lymphangiogenesis in a murine model of breast cancer (25 mg/kg/d p.o.).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank Fontaine et al.
Cell chemical biology, 24(3), 346-359 (2017-02-07)
Pharmacological modulation of transcription factors (TFs) has only met little success over the past four decades. This is mostly due to standard drug discovery approaches centered on blocking protein/DNA binding or interfering with post-translational modifications. Recent advances in the field
Jeroen Overman et al.
eLife, 6 (2017-02-01)
Pharmacological targeting of transcription factors holds great promise for the development of new therapeutics, but strategies based on blockade of DNA binding, nuclear shuttling, or individual protein partner recruitment have yielded limited success to date. Transcription factors typically engage in
Olga Rodak et al.
International journal of molecular sciences, 24(14) (2023-07-29)
The transcription factor SOX18 has been shown to play a crucial role in lung cancer progression and metastasis. In this study, we investigated the effect of Sm4, a SOX18 inhibitor, on cell cycle regulation in non-small cell lung cancer (NSCLC)
Sreeman K Mamidyala et al.
Bioorganic & medicinal chemistry letters, 23(6), 1667-1670 (2013-02-19)
Anacardic acid derivatives exhibit a broad range of biological activities. In this report, an efficient method for the synthesis of anacardic acid derivatives was explored, and a small set of salicylic acid variants synthesised retaining a constant hydrophobic element (a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service