Skip to Content
Merck

Skip To

SML1995

Capzimin dimer

≥96% (HPLC)

Synonym(s):

8,8′-Disulfanediylbis(N-(2-(thiazol-2-yl)ethyl)quinoline-3-carboxamide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 125,00

About This Item

Empirical Formula (Hill Notation):
C30H24N6O2S4
CAS Number:
Molecular Weight:
628.81
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥96% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 125,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥96% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCCC1=NC=CS1)C2=CC3=CC=CC(SSC4=C(N=CC(C(NCCC5=NC=CS5)=O)=C6)C6=CC=C4)=C3N=C2

InChI

1S/C30H24N6O2S4/c37-29(33-9-7-25-31-11-13-39-25)21-15-19-3-1-5-23(27(19)35-17-21)41-42-24-6-2-4-20-16-22(18-36-28(20)24)30(38)34-10-8-26-32-12-14-40-26/h1-6,11-18H,7-10H2,(H,33,37)(H,34,38)

InChI key

RNEOHKZPZKEZCQ-UHFFFAOYSA-N

Biochem/physiol Actions

Capzimin dimer is the stable precursor form of capzimin. Capzimin is a selective inhibitor of proteasome isopeptidase Rpn11, a proteasomal deubiquitinase that is part of the proteosome 19S regulatory particle subunit that removes the polyubiquitin chain from substrates in preparation for degradation. Capzimin was shown to have an IC50 value of 340 nM and have selectivity over related enzymes with 80-fold selectivity for Rpn11 over Csn5, ten-fold over AMSH and six-fold over BRCC36. It stabilized proteasome substrates that would otherwise have been degraded, induced an unfolded protein response, and blocked the proliferation of cancer cells, including some that had become resistant to bortezomib. Capzimin dimer will be quickly reduced to capzimin by glutathione in cell-based systems; DTT should be added to in vitro assays to reduce the disulfide.
Inhibitor of proteasome isopeptidase Rpn11
Capzimin can be used as a therapeutic for cancer treatment.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1016SML2357SML1786
description

≥96% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥96% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear (warmed)

storage condition

desiccated

storage condition

-

storage condition

-

storage condition

-

color

white to beige

color

white to light brown

color

white to beige

color

white to beige


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters