Skip to Content
Merck

Skip To

SML1985

GSK6853

≥98% (HPLC)

Synonym(s):

(R)-N-(1,3-Dimethyl-6-(2-methylpiperazin-1-yl)-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-methoxybenzamide, N-[2,3-Dihydro-1,3-dimethyl-6-[(2R)-2-methyl-1-piperazinyl]-2-oxo-1H-benzimidazol-5-yl]-2-methoxybenzamide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C22H27N5O3
CAS Number:
Molecular Weight:
409.48
UNSPSC Code:
51111800
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -45 to -55°, c = 0.3 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1N(C)C2=CC(N3CCNC[C@H]3C)=C(NC(C4=C(OC)C=CC=C4)=O)C=C2N1C

InChI key

FQWDVNSBYDXPIO-CQSZACIVSA-N

Biochem/physiol Actions

Aqueous soluble, high-affinity, BRPF1 bromodomain-targeting inhibitor with potent cellular efficacy and good pharmacokinetic properties in mice in vivo.
GSK6853 is a GSK5959 structural analog with improved aqueous solubility (140 μg/mL vs. 8 μg/mL for GSK5959 in pH 7.4 PBS), affinity toward BRPF1 bromodomain (pKd = 9.5 vs 8.0 for GSK5959), cellular efficacy (pIC50 = 7.7 vs 6.0 for GSK5959; against NanoLuc-BRPF1 bromodomain interaction with Histone H3.3-HaloTag in HEK293 cells), and selectivity, displaying little or no inhibitory potency against a panel of 48 channels/receptors/enzymes and >1600-fold reduced affinity toward other bromodomains tested (>90-fold in the case of GSK5959), including those of BRPF2 (BRD1) and BRPF3. GSK6853 displays good bioavailability when administered via intraperitoneal injection in mice in vivo (Cmax = 469 ng/mL, Tmax = 0.25 h, F = 85%; 3 mg/kg i.p.). The structural analog GSK9311 serves as a good inactive control (125- and 185-fold reduced potency in cell-free and cell-based assays, respectively). For characterization details of GSK6853, please visit the GSK6853 probe summary on the Structural Genomics Consortium (SGC) website.

GSK9311 is the negative control for the active enantiomer, GSK6853. GSK9311 is available from Sigma. To learn more about and purchase GSK9311, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc

Features and Benefits

GSK6853 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters