Skip to Content
Merck

Skip To

SML1981

NIBR189

≥98% (HPLC), GPR183 antagonist, powder

Synonym(s):

(2E)-3-(4-Bromophenyl)-1-(4-(4-methoxybenzoyl)-1-piperazinyl)-2-propen-1-one, (E)-3-(4-Bromophenyl)-1-(4-(4-methoxybenzoyl)piperazin-1-yl)prop-2-en-1-one, NIBR 189

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 103,00
25 mg
Check Cart for Availability
€ 418,00
€ 355,30

About This Item

Empirical Formula (Hill Notation):
C21H21BrN2O3
CAS Number:
Molecular Weight:
429.31
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 103,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

NIBR189, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

Brc1ccc(cc1)\C=C\C(=O)N2CCN(CC2)C(=O)c3ccc(cc3)OC

InChI

1S/C21H21BrN2O3/c1-27-19-9-5-17(6-10-19)21(26)24-14-12-23(13-15-24)20(25)11-4-16-2-7-18(22)8-3-16/h2-11H,12-15H2,1H3/b11-4+

InChI key

OFHXXBRBGWUOHR-NYYWCZLTSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1601SML2256SML1405
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

room temp

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

DMSO: 10 mg/mL, clear (warmed)

solubility

DMSO: 5 mg/mL, clear (warmed)

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 20 mg/mL, clear

color

white to beige

color

yellow to brown

color

white to beige

color

white to beige

Biochem/physiol Actions

High-affinity, potent and selective EBI2 (GPR183) antagonist with good pharmacokinetic properties and oral availability in mice in vivo.
NBIR189 is a high-affinity, potent and selective EBI2 (GPR183) antagonist (IC50 = 16 nM against 10 nM 7α,25-OHC for binding human EBI2) that inhibits 7α,25-OHC-induced GTPγS binding (IC50 = 8.5 and 7.0 nM, respectively, against 0.33 and 0.1 nM OHC; human EBI2-expressing CHO membrane), calcium mobilization (IC50 = 11 and 15 nM using human or mouse EBI2-transfected cells, respectively), and chemotaxis (IC50 = 0.3 nM against 20 nM OHC-induced U937 migration). NBIR189 displays no inhibitory potency against 5HT2A, muscarinic acetylcholine receptor M2, adrenoreceptor α1A, nor significant affintiy toward 18 other GPCRs/transporters/enzymes, and exhibits good pharmacokinetic properties and oral availability in mice (AUC = 3608 nmol h/L, Cmax = 835 nM, tmax = 1 h, F = 49%; 3 mg/kg p.o.). A useful tool for probing EBI2-mediated physiological functions and autoimmune disorders.

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florian Wanke et al.
Cell reports, 18(5), 1270-1284 (2017-02-02)
Arrival of encephalitogenic T cells at inflammatory foci represents a critical step in development of experimental autoimmune encephalomyelitis (EAE), the animal model for multiple sclerosis. EBI2 and its ligand, 7α,25-OHC, direct immune cell localization in secondary lymphoid organs. CH25H and CYP7B1
Inga Preuss et al.
Biochemical and biophysical research communications, 446(3), 663-668 (2014-02-01)
Oxysterols such as 7 alpha, 25-dihydroxycholesterol (7α,25-OHC) are natural ligands for the Epstein-Barr virus (EBV)-induced gene 2 (EBI2, aka GPR183), a G protein-coupled receptor (GPCR) highly expressed in immune cells and required for adaptive immune responses. Activation of EBI2 by
Aleksandra Rutkowska et al.
Scientific reports, 6, 25520-25520 (2016-05-12)
EBI2 is a G protein-coupled receptor activated by oxysterol 7α, 25-dihydroxycholesterol (7α25HC) and regulates T cell-dependant antibody response and B cell migration. We recently found EBI2 is expressed in human astrocytes, regulates intracellular signalling and modulates astrocyte migration. Here, we
Aurélie S Clottu et al.
Cell reports, 18(1), 213-224 (2017-01-05)
The interaction between oxysterols and the G protein-coupled receptor Epstein-Barr virus-induced gene 2 (EBI2) fine-tunes immune cell migration, a mechanism efficiently targeted by several disease-modifying treatments developed to treat multiple sclerosis (MS), such as natalizumab. We previously showed that memory
Francois Gessier et al.
Journal of medicinal chemistry, 57(8), 3358-3368 (2014-04-01)
Oxysterols have recently been identified as natural ligands for a G protein-coupled receptor called EBI2 (aka GPR183) ( Nature 2011 , 475 , 524 ; 519 ). EBI2 is highly expressed in immune cells ( J. Biol. Chem. 2006

Global Trade Item Number

SKUGTIN
SML1981-25MG04061835302741
SML1981-5MG04061835302758

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service