Skip to Content
Merck
All Photos(1)

Key Documents

SML1975

Sigma-Aldrich

UCB35625

>97% (HPLC)

Synonym(s):

trans-1-(1-Cycloocten-1-ylmethyl)-4-[[(2,7-dichloro-9H-xanthen-9-yl)carbonyl]amino]-1-ethyl-piperidinium iodide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€83.90
25 MG
€214.00

€83.90


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€83.90
25 MG
€214.00

About This Item

Empirical Formula (Hill Notation):
C30H37Cl2IN2O2
CAS Number:
Molecular Weight:
655.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€83.90


Estimated to ship on12 April 2025


Request a Bulk Order

Assay

>97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C30H36Cl2N2O2.HI/c1-2-34(20-21-8-6-4-3-5-7-9-21)16-14-24(15-17-34)33-30(35)29-25-18-22(31)10-12-27(25)36-28-13-11-23(32)19-26(28)29;/h8,10-13,18-19,24,29H,2-7,9,14-17,20H2,1H3;1H/b21-8+;/t24-,34-;

InChI key

FOAFBMYSXIGAOX-MUMZCOSOSA-N

Biochem/physiol Actions

High affinity antagonist for CCR1 and CCR3 receptors, partial agonist at CCR2 and CCR5 receptors
UCB35625 is a high affinity antagonist for CCR1 and CCR3 receptors. Also both UCB35625 and its enantiomer J113863 exhibit low binding affinity toward CCR2 and CCR5 where they activate three Gi and two G0 protein isoforms. UCB35625 does not activate G12 protein isoform. J113863 binding to CCR2 and CCR5 induce the recruitment of b-arrestin 2 whereas UCB35625 did not. J113863 and UCB35625 are biased ligands for the chemokine receptors CCR2 and CCR5.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Filipa Lopes de Mendonça et al.
The Journal of biological chemistry, 280(6), 4808-4816 (2004-11-19)
The chemokine receptor CCR1 and its principal ligand, CCL3/MIP-1alpha, have been implicated in the pathology of several inflammatory diseases including rheumatoid arthritis, multiple sclerosis, and asthma. As such, these molecules are the focus of much research with the ultimate aim
I Sabroe et al.
The Journal of biological chemistry, 275(34), 25985-25992 (2000-06-16)
We describe a small molecule chemokine receptor antagonist, UCB35625 (the trans-isomer J113863 published by Banyu Pharmaceutical Co., patent WO98/04554), which is a potent, selective inhibitor of CCR1 and CCR3. Nanomolar concentrations of UCB35625 were sufficient to inhibit eosinophil shape change
Jenny Corbisier et al.
The Journal of biological chemistry, 292(2), 575-584 (2016-11-30)
Biased agonism at G protein-coupled receptors constitutes a promising area of research for the identification of new therapeutic molecules. In this study we identified two novel biased ligands for the chemokine receptors CCR2 and CCR5 and characterized their functional properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service