Skip to Content
Merck
All Photos(1)

Key Documents

SML1965

Sigma-Aldrich

PKUMDL-WQ-2201

≥98% (HPLC)

Synonym(s):

2-Chloro-4-[5-[[2-[(ethylamino)thioxomethyl]hydrazinylidene]methyl]-2-furanyl]-benzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€86.60
25 MG
€402.00

€86.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€86.60
25 MG
€402.00

About This Item

Empirical Formula (Hill Notation):
C15H14ClN3O3S
CAS Number:
Molecular Weight:
351.81
UNSPSC Code:
12352200
NACRES:
NA.77

€86.60


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(C=CC(C1=CC=C(C=NNC(NCC)=S)O1)=C2)=C2Cl)O

InChI

1S/C15H14ClN3O3S/c1-2-17-15(23)19-18-8-10-4-6-13(22-10)9-3-5-11(14(20)21)12(16)7-9/h3-8H,2H2,1H3,(H,20,21)(H2,17,19,23)

InChI key

XPACBFAEZIPDRT-UHFFFAOYSA-N

Biochem/physiol Actions

PKUMDL-WQ-2201 is a potent and selective non-NAD+-competing allosteric inhibitor of phosphoglycerate dehydrogenase (PHGDH) that selectively inhibits serine synthesis in cancer cells. PKUMDL-WQ-2201 binds to allosteric site II, which is located in the substrate-binding domain. PKUMDL-WQ-2201 inhibits tumor growth of MDA-MB-468 xenografts in mice.
Potent and selective non-NAD+-competing allosteric inhibitor of phosphoglycerate dehydrogenase (PHGDH)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Wang et al.
Cell chemical biology, 24(1), 55-65 (2017-01-04)
Metabolic reprogramming in cancer cells facilitates growth and proliferation. Increased activity of the serine biosynthetic pathway through the enzyme phosphoglycerate dehydrogenase (PHGDH) contributes to tumorigenesis. With a small substrate and a weak binding cofactor, (NAD+), inhibitor development for PHGDH remains

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service