Skip to Content
Merck

Skip To

SML1964

NS3728

≥98% (HPLC)

Synonym(s):

1-[3,5-bis(Trifluoromethyl)phenyl]-3-[4-bromo-2-(1H-tetrazol-5-yl)phenyl]urea, Endovion, N-(3,5-bis(Trifluoromethyl)phenyl)-N′-(4-bromo-2-(1H-tetrazol-5-yl)phenyl)urea, NS 3728, NS-3728

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C16H9BrF6N6O
CAS Number:
Molecular Weight:
495.18
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)NC(=O)Nc2c(ccc(c2)Br)c3n[nH]nn3

InChI

1S/C16H9BrF6N6O/c17-9-1-2-11(13-26-28-29-27-13)12(6-9)25-14(30)24-10-4-7(15(18,19)20)3-8(5-10)16(21,22)23/h1-6H,(H2,24,25,30)(H,26,27,28,29)

InChI key

OQRAKHDEGGGWQO-UHFFFAOYSA-N

Biochem/physiol Actions

A reversible anion channel blocker against VRAC (VSOAC) and CaCC channel anoctamin (ANO).
NS3728 (Endovion) is a di-aryl-urea-based reversible anion channel blocker that is shown to inhibit Ca(2+)-independent Cl- conductance by volume-regulated anion channel (VRAC or VSOAC; IC50 = 400 nM) as well as Ca2+-activated Cl- ccurrent (CaCC) by anoctamin (ANO) in an apparently voltage-dependent manner (IC50 against ANO1 = 2.1 and 0.7 μM at -95 and +50 mV, respectively). NS3728 is reported to be 5.5-times more potent than Tamoxifen against VRAC (IC50 = 0.4 vs. 2.2 μM).


Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters