Skip to Content
Merck

Skip To

SML1963

Abiraterone D4A metabolite

≥98% (HPLC)

Synonym(s):

Δ4-abiraterone, 17-(3-Pyridinyl)-androsta-4,16-dien-3-one, Abiraterone ketone analog, D4-abiraterone, D4A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 459,00

About This Item

Empirical Formula (Hill Notation):
C24H29NO
CAS Number:
Molecular Weight:
347.49
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder

€ 459,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

n1cc(ccc1)C2=CC[C@H]3[C@H]4[C@@H]([C@]5(CCC(=O)C=C5CC4)C)CC[C@@]32C

InChI

1S/C24H29NO/c1-23-11-9-18(26)14-17(23)5-6-19-21-8-7-20(16-4-3-13-25-15-16)24(21,2)12-10-22(19)23/h3-4,7,13-15,19,21-22H,5-6,8-12H2,1-2H3/t19-,21-,22-,23-,24+/m0/s1

InChI key

GYJZZAJJENTSTP-NHFPKVKZSA-N

Biochem/physiol Actions

D4A (Δ4-Abiraterone; 3-Pyridinyl)-androsta-4,16-dien-3-one) is an abiraterone metabolite found to be more active than abiraterone.
D4A (Δ4-Abiraterone; 3-Pyridinyl)-androsta-4,16-dien-3-one) is an abiraterone metabolite found to be more active than abiraterone. Abiraterone is a potent, selective, and orally bioavailable inhibitor of CYP17A1 (CYP450c17), an enzyme that catalyzes two key serial reactions (17 alpha hydroxylase and 17,20 lyase) in androgen and estrogen biosynthesis resulting in the formation of DHEA and androstenedione, which may ultimately be metabolized into testosterone. CYP17 is the key enzyme for androgen biosynthesis in both the testes and adrenals, so its inhibition can stop the production of androgens in both places. It is approved for the treatment of metastatic castration-resistant prostate cancer. D4A blocks not only CYP17A1, but also blocks two other enzymes, 3?-hydroxysteroid dehydrogenase (3?HSD) and steroid-5?-reductase (SRD5A), which are involved in producing the androgen 5?-dihydrotestosterone (DHT), while also blocking the androgen receptor.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML1527SML1642SML2396
description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

description

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 5 mg/mL, clear

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

color

white to beige

color

white to beige

color

white to light brown

color

white to beige

Δ4-abiraterone 3-Pyridinyl compound

-

-

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 1 - Repr. 2 - STOT RE 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters