Skip to Content
Merck
All Photos(1)

Key Documents

SML1943

Sigma-Aldrich

FPMINT

≥98% (HPLC)

Synonym(s):

4-((4-(2-Fluorophenyl)piperazin-1-yl)methyl)-6-imino-N-(naphthalen-1-yl)-1,3,5-triazin-2-amine, 6-[[4-(2-Fluorophenyl)-1-piperazinyl]methyl]-N2-1-naphthalenyl-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€99.50

€99.50


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€99.50

About This Item

Empirical Formula (Hill Notation):
C24H24FN7
CAS Number:
Molecular Weight:
429.49
UNSPSC Code:
12352200
NACRES:
NA.77

€99.50


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(C=CC=C1)=C1N2CCN(CC3=NC(NC4=C(C=CC=C5)C5=CC=C4)=NC(N)=N3)CC2

InChI

1S/C24H24FN7/c25-19-9-3-4-11-21(19)32-14-12-31(13-15-32)16-22-28-23(26)30-24(29-22)27-20-10-5-7-17-6-1-2-8-18(17)20/h1-11H,12-16H2,(H3,26,27,28,29,30)

InChI key

XJZDKGZTSXGLMG-UHFFFAOYSA-N

Biochem/physiol Actions

FPMINT is a potent, irreversible and non-competitive inhibitor of equilibrative nucleoside transporters ENT1 and ENT2. FPMINT is more selective for ENT2 than ENT1.
Potent, irreversible and non-competitive inhibitor of ENT1 and ENT2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philip C T Tang et al.
European journal of pharmacology, 791, 544-551 (2016-10-30)
Equilibrative nucleoside transporters (ENTs) play a crucial role in the transport of nucleoside and nucleoside analogues, which are important for nucleotide synthesis and chemotherapy. In addition, ENTs regulate extracellular adenosine levels in the vicinity of its receptors and hence influence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service