Skip to Content
Merck
All Photos(1)

Key Documents

SML1894

Sigma-Aldrich

SYP-5

≥98% (HPLC)

Synonym(s):

(2E)-1-(5-Hydroxy-2-2-dimethyl-2H-1-benzopyran-6-yl)-3-(2-thienyl)-2-propen-1-one, (E)-1-(5-Hydroxy-2,2-dimethyl-2H-chromen-6-yl)-3-(thio-phen-2-yl)prop-2-en-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€79.30

€79.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€79.30

About This Item

Empirical Formula (Hill Notation):
C18H16O3S
CAS Number:
Molecular Weight:
312.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€79.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C=CC(C)(C)O2)C2=CC=C1C(/C=C/C3=CC=CS3)=O

InChI key

OMVURDVBYIJCOI-FNORWQNLSA-N

Biochem/physiol Actions

SYP-5 is potent and selective inhibitor of HIF-1 activity that decreases HIF-1α and HIF-1β protein levels under hypoxic condition. SYP-5 inhibits tumor cell migration and invasion, as well as tumor angiogenesis.
SYP-5 is potent and selective inhibitor of HIF-1 activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Hui Wang et al.
European journal of pharmacology, 791, 560-568 (2016-10-30)
Hypoxia-inducible factor-1 (HIF-1) plays an essential role in carcinogenesis. The overexpression of HIF-1 induced by hypoxia is closely associated with metastasis, poor prognosis and high mortality. In this study, a novel HIF-1 inhibitor SYP-5 was first observed by the luciferase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service