Skip to Content
Merck

Skip To

SML1772

Givinostat hydrochloride hydrate

≥95% (HPLC), HDAC inhibitor, powder

Synonym(s):

Diethyl-[6-(4-hydroxycarbamoyl-phenyl carbamoyloxymethyl)-naphthalen-2-yl methyl]-ammonium chloride, ITF2357, N-[4-[(Hydroxyamino)carbonyl]phenyl]-carbamic acid, [6-[(diethylamino)methyl]-2-naphthalenyl]methyl ester hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
€ 142,00
25 mg
Check Cart for Availability
€ 571,00

About This Item

Empirical Formula (Hill Notation):
C24H27N3O4 · HCl · H2O
CAS Number:
Molecular Weight:
475.97
NACRES:
NA.77
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated

€ 142,00


Check Cart for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Givinostat hydrochloride hydrate, ≥95% (HPLC)

Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[H]Cl.[H]O[H].CCN(CC)CC1=CC=C(C=C(COC(NC2=CC=C(C(NO)=O)C=C2)=O)C=C3)C3=C1

InChI

1S/C24H27N3O4.ClH.H2O/c1-3-27(4-2)15-17-5-7-21-14-18(6-8-20(21)13-17)16-31-24(29)25-22-11-9-19(10-12-22)23(28)26-30;;/h5-14,30H,3-4,15-16H2,1-2H3,(H,25,29)(H,26,28);1H;1H2

InChI key

FKGKZBBDJSKCIS-UHFFFAOYSA-N

Application

Givinostat hydrochloride hydrate has been used as a histone deacetylase inhibitor to test its effect on the human immunodeficiency virus reactivation in CD4+ T-cell model.

Biochem/physiol Actions

Givinostat (ITF2357) is a hydroxamate HDAC inhibitor that inhibits class I and class II enzymes.
Givinostat (ITF2357) is a hydroxamate HDAC inhibitor that inhibits class I and class II enzymes. Givinostat posseses a very promising activity profile in multiple myeloma and acute myelogenous leukemia in vitro and in vivo. Givinostat also has anti-inflammatory activity and inhibits the secretion of the tumor necrosis factor-alpha (TNF-α), IL-1, and IL-6. Givinostat has been in multiple Phase 2 studies for both inflammatory diseases (Duchenne Muscular Dystrophy , Juvenile arthritis, Polycythemia Vera) and blood cancers (myelomas and lymphomas).
Givinostat is known to mediate the reduction of the human immunodeficiency virus release in macrophages.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters