Skip to Content
Merck
All Photos(1)

Key Documents

SML1770

Sigma-Aldrich

PhiKan083 hydrochloride

≥98% (HPLC)

Synonym(s):

9-Ethyl-N-methyl-9H-Carbazole-3-methanamine hydrochloride, PK-083 hydrochloride, PK083 hydrochloride, PhiKan-083 hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€96.10
25 MG
€388.00

€96.10


Estimated to ship on23 May 2025



Select a Size

Change View
5 MG
€96.10
25 MG
€388.00

About This Item

Empirical Formula (Hill Notation):
C16H18N2 · HCl
CAS Number:
Molecular Weight:
274.79
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€96.10


Estimated to ship on23 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNCC1=CC2=C(C=C1)N(CC)C3=C2C=CC=C3.[H]Cl

InChI key

IXWVUPURFWFRNE-UHFFFAOYSA-N

Biochem/physiol Actions

Mutant p53 stabilizer
PhiKan083 is a small molecule that binds within a cavity on the surface of Y220C-mutated p53. This mutation, which is not involved in DNA interaction, causes a destabilization of the protein. PhiKan083 binding acts to stabilize the mutant p53 and slow the rate of denaturation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias R Bauer et al.
ACS chemical biology, 11(8), 2265-2274 (2016-06-09)
Many oncogenic mutants of the tumor suppressor p53 are conformationally unstable, including the frequently occurring Y220C mutant. We have previously developed several small-molecule stabilizers of this mutant. One of these molecules, PhiKan083, 1-(9-ethyl-9H-carbazole-3-yl)-N-methylmethanamine, binds to a mutation-induced surface crevice with
Frank M Boeckler et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(30), 10360-10365 (2008-07-25)
The tumor suppressor p53 is mutationally inactivated in approximately 50% of human cancers. Approximately one-third of the mutations lower the melting temperature of the protein, leading to its rapid denaturation. Small molecules that bind to those mutants and stabilize them
Joana Soares et al.
Oncotarget, 7(4), 4326-4343 (2016-01-07)
Restoration of the p53 pathway, namely by reactivation of mutant (mut) p53, represents a valuable anticancer strategy. Herein, we report the identification of the enantiopure tryptophanol-derived oxazoloisoindolinone SLMP53-1 as a novel reactivator of wild-type (wt) and mut p53, using a
Keelan Z Guiley et al.
Cancer discovery, 13(1), 56-69 (2022-10-06)
The transcription factor and tumor suppressor protein p53 is the most frequently mutated and inactivated gene in cancer. Mutations in p53 result in deregulated cell proliferation and genomic instability, both hallmarks of cancer. There are currently no therapies available that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service