Skip to Content
Merck
All Photos(1)

Key Documents

SML1733

Sigma-Aldrich

FINDY

≥98% (HPLC)

Synonym(s):

(5Z)-5-[[4-Methoxy-3-[2-(trimethylsilyl)ethynyl]phenyl]methylene]-2-thioxo-4-thiazolidinone, (Z)-5-(4-Methoxy-3-((trimethylsilyl)ethynyl)benzylidene)-2-thioxothiazolidin-4-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€108.00
25 MG
€440.00

€108.00


Estimated to ship on27 April 2025



Select a Size

Change View
5 MG
€108.00
25 MG
€440.00

About This Item

Empirical Formula (Hill Notation):
C16H17NO2S2Si
CAS Number:
Molecular Weight:
347.53
UNSPSC Code:
12352200
NACRES:
NA.77

€108.00


Estimated to ship on27 April 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[Si](C)(C)C#CC1=C(OC)C=CC(/C=C2SC(NC\2=O)=S)=C1

InChI

1S/C16H17NO2S2Si/c1-19-13-6-5-11(9-12(13)7-8-22(2,3)4)10-14-15(18)17-16(20)21-14/h5-6,9-10H,1-4H3,(H,17,18,20)/b14-10-

InChI key

JHFDWHHUUUMRLK-UVTDQMKNSA-N

Biochem/physiol Actions

A thioxothiazolidinone that specifically targets newly synthesized cellular DYRK1A, but not DYRK1B or DYRK2, for proteasomal degradation.
FINDY is a cell-permeable thioxothiazolidinone derivative that specifically targets newly synthesized cellular DYRK1A, but not DYRK1B or DYRK2, for proteasomal degradation by suppressing DYKR1A intramolecular Ser97 autophosphorylation, a necessary event for folding/maturation of newly translated DYRK1A. Unlike INDY and other ATP-competitive DYRK inhibitors, FINDY is ineffective against the kinase activity of DYRK1A, DYRK1B, DYRK2, or DYRK3 (IC50 >10 μM) and inhibits only five kinases (GSK3β, MARK4, PIM1, PIM3, PLK3) by over 75% at 10 μM among a panel of 271 other kinases. FINDY (2.5 μM) is shown to selectively rescue the developmental defect of Xenopus embryos induced by the overexpression of DYRK1A, but not DYRK1B, while INDY prodrug (2.5 μM) treatment indiscriminately prevents defect caused by both DYRK1A and DYRK1B expression.
FINDY is a structural analog of RD0392, a canonical ATP-competitive inhibitor[1] of mature dual-specificity tyrosine phosphorylation-regulated kinase 1A (DYRK1A).[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Umezawa et al.
Molecules (Basel, Switzerland), 26(3) (2021-01-31)
Drug discovery using small molecule inhibitors is reaching a stalemate due to low selectivity, adverse off-target effects and inevitable failures in clinical trials. Conventional chemical screening methods may miss potent small molecules because of their use of simple but outdated
Isao Kii et al.
Nature communications, 7, 11391-11391 (2016-04-23)
Autophosphorylation of amino-acid residues is part of the folding process of various protein kinases. Conventional chemical screening of mature kinases has missed inhibitors that selectively interfere with the folding process. Here we report a cell-based assay that evaluates inhibition of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service