Skip to Content
Merck
All Photos(1)

Key Documents

SML1728

Sigma-Aldrich

STK16-IN-1

≥98% (HPLC)

Synonym(s):

1-(4-Fluoro-3-methylphenyl)-1,7-dihydro-2H-pyrrolo[2,3-h]-1,6-naphthyridin-2-one, 1-(4-Fluoro-3-methylphenyl)-1H-pyrrolo[2,3-h][1,6]naphthyridin-2(7H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€91.80

€91.80


Estimated to ship on31 May 2025



Select a Size

Change View
5 MG
€91.80

About This Item

Empirical Formula (Hill Notation):
C17H12FN3O
CAS Number:
Molecular Weight:
293.30
UNSPSC Code:
12352202
NACRES:
NA.77

€91.80


Estimated to ship on31 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CC1=CC(N2C(C(C=CN3)=C3N=C4)=C4C=CC2=O)=CC=C1F

InChI

1S/C17H12FN3O/c1-10-8-12(3-4-14(10)18)21-15(22)5-2-11-9-20-17-13(16(11)21)6-7-19-17/h2-9H,1H3,(H,19,20)

InChI key

WQNRDXHKVSKUPI-UHFFFAOYSA-N

Biochem/physiol Actions

STK16-IN-1 is a cell-permeable pyrrolonaphthyridinone compound that acts as an ATP-competitive inhibitor against NAK (Numb Associated Kinase) family serine/threonine kinase STK16 (IC50/substrate = 0.91 μM/4EBP1 and 1.2 μM/DRG1, [ATP] = 20 μM) with excellent selectivity over a panel of 378 kinases. Despite its inhibitory potency against PI 3-Ks (IC50 = 1.07, 0.856, 0.867 μM against p110α/p85α, p110σ/p85α, p110γ , respectively) in cell-free assays, STK16-IN-1 is ineffective against their cellular activity up to 10 μM concentration. STK16-IN-1 treatment (10 μM for 72 hrs) is shown to cause cell number reduction (~50% in HCT-116 and MCF-7 cultures), as well as increased populations of binucleated cells (~5-fold in MCF-7 culture) and G2/M cells (~7 and 3-fold in HCT-116 and MCF-7 cultures, respectively) in an STK16-dependent manner. A useful tool for elucidating STK16 biological functions.
STK16-IN-1 is a cell-permeable pyrrolonaphthyridinone compound that acts as an ATP-competitive inhibitor against NAK family serine/threonine kinase STK16 with excellent selectivity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service