Skip to Content
Merck
All Photos(1)

Key Documents

SML1705

Sigma-Aldrich

Cbz-B3A

≥98% (HPLC)

Synonym(s):

(8S)-3,8-Bis[[(1,1-dimethylethoxy)carbonyl]amino]-9-oxo-2,4,10,17-tetraazaoctadec-3-enedioic acid 1-(1,1-dimethylethyl) 18-(phenylmethyl) ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€127.00

€127.00


Check Cart for Availability


Select a Size

Change View
5 MG
€127.00

About This Item

Empirical Formula (Hill Notation):
C35H58N6O9
CAS Number:
Molecular Weight:
706.87
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€127.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(OCC1=CC=CC=C1)NCCCCCCNC([C@H](CCCN/C(NC(OC(C)(C)C)=O)=N\C(OC(C)(C)C)=O)NC(OC(C)(C)C)=O)=O

InChI

1S/C35H58N6O9/c1-33(2,3)48-30(44)39-26(20-17-23-37-28(40-31(45)49-34(4,5)6)41-32(46)50-35(7,8)9)27(42)36-21-15-10-11-16-22-38-29(43)47-24-25-18-13-12-14-19-25/h12-14,18-19,26H,10-11,15-17,20-24H2,1-9H3,(H,36,42)(H,38,43)(H,39,44)(H2,37,40,41,45,46)/t26-/m

InChI key

JGEWUYSTHGIDHO-SANMLTNESA-N

Biochem/physiol Actions

Cbz-B3A is a potent and selective inhibitor of mTORC1 signaling that inhibits the phosphorylation of eIF4E binding protein 1 (4EBP1) and blocks 68% of translation. Cbz-B3A binds to ubiquilins 1, 2, and 4, which prevents mTORC1 activation.
Cbz-B3A is a potent and selective inhibitor of mTORC1 signaling.

Other Notes

Light and air sensitive.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service