Skip to Content
Merck
All Photos(1)

Key Documents

SML1688

Sigma-Aldrich

KT172

≥98% (HPLC)

Synonym(s):

(2-Benzylpiperidin-1-yl)(4-(2′-methoxy-[1,1′-biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl)methanone, Cravatt Reagent, [4-(2′-Methoxy[1,1′-biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl][2-(phenylmethyl)-1-piperidinyl]-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€102.00
25 MG
€300.00

€102.00


Check Cart for Availability


Select a Size

Change View
5 MG
€102.00
25 MG
€300.00

About This Item

Empirical Formula (Hill Notation):
C28H28N4O2
CAS Number:
Molecular Weight:
452.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€102.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1N=NC(C2=CC=C(C3=C(OC)C=CC=C3)C=C2)=C1)N4CCCCC4CC5=CC=CC=C5

InChI

1S/C28H28N4O2/c1-34-27-13-6-5-12-25(27)22-14-16-23(17-15-22)26-20-32(30-29-26)28(33)31-18-8-7-11-24(31)19-21-9-3-2-4-10-21/h2-6,9-10,12-17,20,24H,7-8,11,18-19H2,1H3

InChI key

MXVVJNJNDCJCFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

KT172 is a Diacylglycerol lipase (DAGL) inhibitor selective for DAGLβ.
KT172 is a Diacylglycerol lipase (DAGL) inhibitor selective for DAGLβ. Diacylglycerol lipases DAGLα and DAGLβ are serine hydrolases that biosynthesize the endocannabinoid 2-arachidonoylglycerol (2-AG). A lack of selective inhibitors has hampered study of these lipases. KT172 is a new potent and selective DAGLβ inhibitor with an IC50 of 60 nM for recombinant DAGLβ vs 140 nM for DAGLα, although it is not active against DAGLα in vivo, and negligible activity against FAAH, MGLL and ABHD11, other key enzymes involved in endocannabinoid signaling or against cytosolic phospholipase A2 (cPLA2 or PLA2G4A). The main off target inhibition against ABHD6 (IC50 values of 5 nM) can be controlled for by use of the related compounds, KT195 SML1308 or KT185 SML1249, potent and selective ABDH6 inhibitors with negligible activity against DAGLβ. KT172 disrupts the lipid network involved in macrophage inflammatory responses, lowering 2-AG, as well as arachidonic acid and eicosanoids, in mouse peritoneal macrophages.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service