Skip to Content
Merck
All Photos(1)

Key Documents

SML1613

Sigma-Aldrich

CC-401 dihydrochloride

≥95% (HPLC)

Synonym(s):

1-[5-(1H-1,2,4-Triazol-5-yl)-1H-indazol-3-yl]-3-(2-piperidinoethoxy)benzene dihydrochloride, 3-(3-(2-(Piperidin-1-yl)ethoxy)phenyl)-5-(1H-1,2,4-triazol-3-yl)-1H-indazole dihydrochloride, 3-[3-[2-(1-Piperidinyl)ethoxy]phenyl]-5-(1H-1,2,4-triazol-5-yl)-1H-iIndazole dihydrochloride, CC 401, CC401, CID 10430360

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€198.00

€198.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€198.00

About This Item

Empirical Formula (Hill Notation):
C22H24N6O · 2HCl
CAS Number:
Molecular Weight:
461.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€198.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[H]Cl.C1(C2=NC=NN2)=CC3=C(NN=C3C4=CC=CC(OCCN5CCCCC5)=C4)C=C1

InChI

1S/C22H24N6O.ClH/c1-2-9-28(10-3-1)11-12-29-18-6-4-5-16(13-18)21-19-14-17(22-23-15-24-27-22)7-8-20(19)25-26-21;/h4-8,13-15H,1-3,9-12H2,(H,25,26)(H,23,24,27);1H

InChI key

OIBVXKYKWOUGAO-UHFFFAOYSA-N

Biochem/physiol Actions

CC-401 is a cell penetrant, potent and selective c-Jun N terminal kinase (JNK) inhibitor that blocks JNK signaling in the rat obstructed kidney and inhibits renal fibrosis in the unilateral ureteral obstruction model. CC-401 in combination with chemotherapy exhibits synergism in colon cancer cell lines.
CC-401 is a cell penetrant, potent and selective c-Jun N terminal kinase (JNK) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service