Skip to Content
Merck
All Photos(1)

Key Documents

SML1610

Sigma-Aldrich

Dapiprazole hydrochloride

≥98% (HPLC)

Synonym(s):

5,6,7,8-Tetrahydro-3-[2-[4-(2-methylphenyl)-1-piperazinyl]ethyl]-1,2,4-triazolo[4,3-a]pyridine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€102.00
50 MG
€412.00

€102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€102.00
50 MG
€412.00

About This Item

Empirical Formula (Hill Notation):
C19H27N5 · HCl
CAS Number:
Molecular Weight:
361.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€102.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=CC=C1N2CCN(CCC3=NN=C4N3CCCC4)CC2.[H]Cl

InChI

1S/C19H27N5.ClH/c1-16-6-2-3-7-17(16)23-14-12-22(13-15-23)11-9-19-21-20-18-8-4-5-10-24(18)19;/h2-3,6-7H,4-5,8-15H2,1H3;1H

InChI key

ZIODNPFQZIHCOE-UHFFFAOYSA-N

Gene Information

human ... ADRA1A(148)

Biochem/physiol Actions

Dapiprazole hydrochloride is an an α-adrenergic blocking agent that is used to reverse mydriasis after eye examination.
Dapiprazole hydrochloride is an an α-adrenergic blocking agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service