Skip to Content
Merck
All Photos(1)

Key Documents

SML1590

Sigma-Aldrich

Alcaftadine

≥98% (HPLC)

Synonym(s):

6,11-Dihydro-11-(1-methyl-4-piperidinylidene)-5H-imidazo[2,1-b][3]benzazepine-3-carboxaldehyde, R 89674

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€69.40

€69.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€69.40

About This Item

Empirical Formula (Hill Notation):
C19H21N3O
CAS Number:
Molecular Weight:
307.39
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€69.40


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1CCC(CC1)=C2C3=CC=CC=C3CCN4C2=NC=C4C=O

InChI

1S/C19H21N3O/c1-21-9-6-15(7-10-21)18-17-5-3-2-4-14(17)8-11-22-16(13-23)12-20-19(18)22/h2-5,12-13H,6-11H2,1H3

InChI key

MWTBKTRZPHJQLH-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Biochem/physiol Actions

Alcaftadine is a potent inverse agonist at H1, H2 and H4 histamine receptors.
Alcaftadine is a potent inverse agonist at H1, H2 and H4 histamine receptors. Alcaftadine is antiallergic agent that exhibit anti-inflammatory and mast cell stabilizing effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service