Skip to Content
Merck
All Photos(1)

Key Documents

SML1526

Sigma-Aldrich

Eltoprazine hydrochloride

≥98% (HPLC)

Synonym(s):

1-(2,3-Dihydro-1,4-benzodioxin-5-yl)-piperazine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€361.00

€361.00


Check Cart for Availability


Select a Size

Change View
25 MG
€361.00

About This Item

Empirical Formula (Hill Notation):
C12H16N2O2 · HCl
CAS Number:
Molecular Weight:
256.73
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€361.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

room temp

SMILES string

[H]Cl.C1(OCCO2)=C2C(N3CCNCC3)=CC=C1

InChI

1S/C12H16N2O2.ClH/c1-2-10(14-6-4-13-5-7-14)12-11(3-1)15-8-9-16-12;/h1-3,13H,4-9H2;1H

InChI key

JFSOSUNPIXJCIX-UHFFFAOYSA-N

General description

Eltoprazine, a serotonergic agent, possesses specific anti-aggressive (serenic) properties in various animal models without triggering sedation, motor disturbances, or muscle relaxation.

Biochem/physiol Actions

Eltoprazine hyrochloride is a serotonin 5-HT1A/1B receptor agonist that has been shown to reduce L-DOPA-induced dyskinesias in in experimental models of Parkinson′s Disease and in human patients. It has Ki values for 5-HT1A, 5-HT1B and 5-HT1C receptors of 40 nM and 52 nM, respectively.
Serotonin 5-HT1A/1B receptor agonist

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service