Skip to Content
Merck
All Photos(1)

Key Documents

SML1474

Sigma-Aldrich

Oxiconazole nitrate

≥98% (HPLC)

Synonym(s):

1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)-ethanone (1Z)-O-[(2,4-dichlorophenyl)methyl]oxime nitrate, 2′,4′-Dichloro-2-imidazol-1-ylacetophenone (Z)-[0-(2,4-dichlorobenzyl)oxime] nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€104.00
50 MG
€284.00

€104.00


Estimated to ship on29 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€104.00
50 MG
€284.00

About This Item

Empirical Formula (Hill Notation):
C18H13Cl4N3O · HNO3
CAS Number:
Molecular Weight:
492.14
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

€104.00


Estimated to ship on29 April 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C18H13Cl4N3O.HNO3/c19-13-2-1-12(16(21)7-13)10-26-24-18(9-25-6-5-23-11-25)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11H,9-10H2;(H,2,3,4)/b24-18+;

InChI key

WVNOAGNOIPTWPT-NDUABGMUSA-N

Biochem/physiol Actions

Oxiconazole is a broad-spectrum antifungal agent that inhibits ergosterol biosynthesis.
Oxiconazole is a broad-spectrum antifungal agent that inhibits ergosterol biosynthesis. Oxiconazole destabilizes Lanosterol 14-α demethylase (fungal cyctochrome P450 51).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service