Skip to Content
Merck
All Photos(1)

Key Documents

SML1444

Sigma-Aldrich

(±)-Mepivacaine hydrochloride

≥98% (HPLC)

Synonym(s):

1-Methyl-2,6-pipecoloxylidine hydrochloride, Carbocaine hydrochloride, N-(2,6-Dimethylphenyl)-1-methyl-2-piperidinecarboxamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€179.00

€179.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€179.00

About This Item

Empirical Formula (Hill Notation):
C15H22N2O · HCl
CAS Number:
Molecular Weight:
282.81
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€179.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

room temp

SMILES string

[Cl-].N1(C(CCCC1)C(=O)Nc2c(cccc2C)C)C.[H+]

InChI

1S/C15H22N2O.ClH/c1-11-7-6-8-12(2)14(11)16-15(18)13-9-4-5-10-17(13)3;/h6-8,13H,4-5,9-10H2,1-3H3,(H,16,18);1H

InChI key

RETIMRUQNCDCQB-UHFFFAOYSA-N

Biochem/physiol Actions

(±)-Mepivacaine hydrochloride is a local anesthetic that reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current.
(±)-Mepivacaine hydrochloride is a local anesthetic that reversibly blocks transient Na+ inward current, as well as the steady-state K+ outward current. (±)-Mepivacaine blocks tandem pore (TASK) and Kv1.5 potassium channels in model systems.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service