Skip to Content
Merck
All Photos(1)

Key Documents

SML1442

Sigma-Aldrich

(Z-LL)2 Ketone

≥98% (HPLC)

Synonym(s):

(Z-Leu-Leu-NHCH2)2CO, 1,3-Di-(N-Carboxybenzoyl-L-leucyl-L-leucyl)amino Acetone, 2,2′-(2-Oxo-1,3-propanediyl)bis[N-[(phenylmethoxy)carbonyl]-L-leucyl-L-leucinamide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€63.30

€63.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€63.30

About This Item

Empirical Formula (Hill Notation):
C43H64N6O9
CAS Number:
Molecular Weight:
809.00
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€63.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light yellow

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(N[C@@H](CC(C)C)C(N[C@@H](CC(C)C)C(NCC(CNC([C@H](CC(C)C)NC([C@H](CC(C)C)NC(OCC1=CC=CC=C1)=O)=O)=O)=O)=O)=O)OCC2=CC=CC=C2

InChI

1S/C43H64N6O9/c1-27(2)19-34(46-40(53)36(21-29(5)6)48-42(55)57-25-31-15-11-9-12-16-31)38(51)44-23-33(50)24-45-39(52)35(20-28(3)4)47-41(54)37(22-30(7)8)49-43(56)58-26-32-17-13-10-14-18-32/h9-18,27-30,34-37H,19-26H2,1-8H3,(H,44,51)(H,45,52)(H,46,53)(H,47,54)(H,48,55)(H,49,56)/t34-,35-,36-,37-/m0/s1

InChI key

JTVCWQUNPMKMER-BQYLNSIHSA-N

Biochem/physiol Actions

(Z-LL)2 Ketone is a selective inhibitor of signal peptide peptidase (SPP) that inhibits processing of the p-Prl signal peptide (IC50 = ~ 50 nM) without affecting the activities of signal peptidases and other proteases such as lysosomal cathepsins and proteasomes.
(Z-LL)2 Ketone is a selective inhibitor of signal peptide peptidase (SPP).
(Z-LL)2 Ketone serves as an inverse γ-secretase modulator (iGSM) by changing processivity of γ-secretase cleavage.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential Inhibition of Signal Peptide Peptidase Family Members by Established gamma-Secretase Inhibitors
Ran Y, et al.
PLoS ONE, 10 (2015)
Differential Inhibition of Signal Peptide Peptidase Family Members by Established gamma-Secretase Inhibitors
Ran Y, et al.
Testing, 10 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service