Skip to Content
Merck
All Photos(1)

Key Documents

SML1401

Sigma-Aldrich

XL413 hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-8-Chloro-2-(pyrrolidin-2-yl)benzofuro[3,2-d]pyrimidin-4(3H)-one hydrochloride, BMS-863233, XL-413

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€190.00
25 MG
€628.00

€190.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€190.00
25 MG
€628.00

About This Item

Empirical Formula (Hill Notation):
C14H12ClN3O2 · xHCl
CAS Number:
Molecular Weight:
289.72 (free base basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€190.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CC2=C(C=C1)OC3=C2N=C([C@H]4NCCC4)NC3=O.C

InChI

1S/C14H12ClN3O2.CH4/c15-7-3-4-10-8(6-7)11-12(20-10)14(19)18-13(17-11)9-2-1-5-16-9;/h3-4,6,9,16H,1-2,5H2,(H,17,18,19);1H4/t9-;/m0./s1

InChI key

UYBMDYGAVMHMGF-FVGYRXGTSA-N

Biochem/physiol Actions

XL413 is a selective inhibitor of the serine-threonine kinase CDC7 (cell division cycle 7 homolog), a regulator of cell cycle checkpoint control that has been implicated in protecting tumor cells from apoptotic cell death during replication stress. XL413 has an IC50 of 3.4 nM for CDC7, compared to 215 nM, 42 nM and 18 nM for CK2, PIM1 and pMCM2, respectively. XL413 caused significant tumor growth regression in rodent models, causing cell cycle arrest in the late S to G2 phase.
XL413 is a selective inhibitor of the serine-threonine kinase CDC7.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuichiro Saito et al.
Journal of cell science, 133(12) (2020-05-22)
RIF1 controls both DNA replication timing and the DNA double-strand break (DSB) repair pathway to maintain genome integrity. However, it remains unclear how RIF1 links these two processes following exposure to ionizing radiation (IR). Here, we show that inhibition of
Mehdi Bouhaddou et al.
Cell, 182(3), 685-712 (2020-07-10)
The causative agent of the coronavirus disease 2019 (COVID-19) pandemic, severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), has infected millions and killed hundreds of thousands of people worldwide, highlighting an urgent need to develop antiviral therapies. Here we present a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service