Skip to Content
Merck
All Photos(1)

Key Documents

SML1394

Sigma-Aldrich

Olmesartan

≥98% (HPLC)

Synonym(s):

4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid, Olmesartan acid

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€138.00

€138.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
€138.00

About This Item

Empirical Formula (Hill Notation):
C24H26N6O3
CAS Number:
Molecular Weight:
446.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€138.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C)(C)C1=C(C(O)=O)N(CC2=CC=C(C3=C(C4=NNN=N4)C=CC=C3)C=C2)C(CCC)=N1

InChI

1S/C24H26N6O3/c1-4-7-19-25-21(24(2,3)33)20(23(31)32)30(19)14-15-10-12-16(13-11-15)17-8-5-6-9-18(17)22-26-28-29-27-22/h5-6,8-13,33H,4,7,14H2,1-3H3,(H,31,32)(H,26,27,28,29)

InChI key

VTRAEEWXHOVJFV-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Application

Olmesartan has been used as an angiotensin II receptor 1 (AT1) receptor inhibitor to study the effect of angiotensin II type 1 receptor interactions in the regulation of renal afferent arterioles in angiotensin II-dependent hypertension.[1]

Biochem/physiol Actions

Olmesartan is an Angiotensin II Type I receptor blocker.
Olmesartan is an Angiotensin II Type I receptor blocker. Olmesartan is the active form of the antihypertensive drug olmesartan medoxomil.
Olmesartan possesses anti-inflammatory and anti-oxidative stress properties. It protects neuronal cells against oligomerized amyloid β (Aβ)-induced cellular senescence.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elham Bakhtiari et al.
Toxicology mechanisms and methods, 25(8), 614-621 (2015-09-04)
Over expression of renin-angiotensin system (RAS) and nuclear factor-kappaB (NF-κB) has a major role in many cancers. It has been suggested that some angiotensin receptor blockers (ARBs) could reduce the proliferation of cancer cells. The role of NF-κB pathway has
C Custodero et al.
Ageing research reviews, 46, 42-59 (2018-05-29)
Growing evidence suggests chronic low-grade inflammation (LGI) as a possible mechanism underlying the aging process. Some biological and pharmaceutical compounds may reduce systemic inflammation and potentially avert functional decline occurring with aging. The aim of the present meta-analysis was to
Bapi Gorain et al.
Regulatory toxicology and pharmacology : RTP, 82, 20-31 (2016-11-07)
Poor aqueous solubility and unfavourable de-esterification of olmesartan medoxomil (a selective angiotensin II receptor blocker), results in low oral bioavailability of less than 26%. Improvement of oral bioavailability with prolonged pharmacodynamics activity of olmesartan in Wistar rats had been approached
Mi-Yeon Yu et al.
PloS one, 13(8), e0202676-e0202676 (2018-08-28)
Albuminuria is a predictor of disease progression in patients with chronic kidney disease (CKD). However, the ability of proteinuria parameters measured at various time periods to predict renal outcomes is unclear. This observational cohort study included 165 non-diabetic hypertensive CKD
Nanako Muraya et al.
Oxidative medicine and cellular longevity, 2018, 7635274-7635274 (2018-07-04)
Oxidative stress induced by hyperuricemia is closely associated with the renin-angiotensin system, as well as the onset and progression of cardiovascular disease (CVD) and chronic kidney disease (CKD). It is therefore important to reduce oxidative stress to treat hyperuricemia. We

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service