Skip to Content
Merck
All Photos(1)

Key Documents

SML1295

Sigma-Aldrich

RBC8

≥98% (HPLC)

Synonym(s):

6-Amino-4-(2,5-dimethoxyphenyl)-1,4-dihydro-3-(2-naphthalenyl)-pyrano[2,3-c]pyrazole-5-carbonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€290.00

€112.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€290.00

About This Item

Empirical Formula (Hill Notation):
C25H20N4O3
CAS Number:
Molecular Weight:
424.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€112.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

NC1=C(C#N)C(C2=C(OC)C=CC(OC)=C2)C(C(C3=CC=C(C=CC=C4)C4=C3)=NN5)=C5O1

InChI

1S/C25H20N4O3/c1-30-17-9-10-20(31-2)18(12-17)21-19(13-26)24(27)32-25-22(21)23(28-29-25)16-8-7-14-5-3-4-6-15(14)11-16/h3-12,21H,27H2,1-2H3,(H,28,29)

InChI key

CLMQBVUFKIKYLU-UHFFFAOYSA-N

Biochem/physiol Actions

RBC8 is a selective inhibitor of the Ras-like GTPases RalA and RalB, downstream mediators of Ras signalling, without direct inihibition of Ras or RhoA activity. RBC8 binds to a site on the GDP-bound form of Ral, its inactive state, inhibiting the binding of Ral to its effectors, such as Ral-binding protein 1 (RALBP1; also known as RLIP760, which are involved in proliferation, survival, and metastasis of several human cancers. RBC8 inhibited growth in human luung cancer cell lines with IC50 values of 2.0 μM in H2122 cells and 1.3 μM in H358 cells, and also inihbited tumor xenograft growth..
RBC8 is a selective inhibitor of the Ras-like GTPases RalA and RalB.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service