Skip to Content
Merck
All Photos(1)

Key Documents

SML1234

Sigma-Aldrich

HAMNO

≥98% (HPLC)

Synonym(s):

(1Z)-1-[(2-Hydroxyanilino)methylidene]naphthalen-2-one, 1-[[(2-Hydroxyphenyl)imino]methyl]-2-naphthalenol, CID 6335338, MLS000737724, NS C111847, NSC111847

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€438.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€438.00

About This Item

Empirical Formula (Hill Notation):
C17H13NO2
CAS Number:
Molecular Weight:
263.29
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c3c(cccc3)O)\C=C1\c2c(cccc2)C=CC\1=O

InChI key

NADCEWZYITXLKJ-KAMYIIQDSA-N

Biochem/physiol Actions

HAMNO is a potent and selective inhibitor of replication protein A (RPA) interactions with proteins involved in the replication stress response. HAMNO binds the N-terminal domain of RPA70 and inhibits both ATR autophosphorylation and phosphorylation of RPA32 Ser33 by ATR. HAMNO enhances DNA replication stress in cancer cells and slow tumor growth in vivo.
HAMNO is a potent and selective inhibitor of replication protein A (RPA) interactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service