Skip to Content
Merck
All Photos(1)

Key Documents

SML1217

Sigma-Aldrich

TG4-155

≥98% (HPLC)

Synonym(s):

(2E)-N-[2-(2-Methyl-1H-indol-1-yl)ethyl]-3-(3,4,5-trimethoxyphenyl)-2-propenamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€112.00
25 MG
€438.00

€112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€112.00
25 MG
€438.00

About This Item

Empirical Formula (Hill Notation):
C23H26N2O4
CAS Number:
Molecular Weight:
394.46
UNSPSC Code:
12352200
NACRES:
NA.77

€112.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]2(c3c(cc2C)cccc3)CCNC(=O)\C=C\c1cc(c(c(c1)OC)OC)OC

InChI

1S/C23H26N2O4/c1-16-13-18-7-5-6-8-19(18)25(16)12-11-24-22(26)10-9-17-14-20(27-2)23(29-4)21(15-17)28-3/h5-10,13-15H,11-12H2,1-4H3,(H,24,26)/b10-9+

InChI key

YBHUXHFZLMFETJ-MDZDMXLPSA-N

Biochem/physiol Actions

TG4-155 is a brain penetrant prostaglandin EP2 specific antagonist.
TG4-155 is a brain penetrant prostaglandin EP2 specific antagonist. TG4-155 has a 4730X selectivity over EP4 (2.4 nM vs 11.4 mM in binding assays), and a 7 fold selectivity over DP1 receptors. TG4-155 crosses the blood brain barrier, and has no antagonistic effects on EP4 signaling, and displays neuroprotective effects in a pilocarpine-induced status epilepticus (SE) seizure model. TG4-155 does not interfere with G-protein function or phosphodiesterase activity, and does not inhibit COX-1 or COX-2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiange Qiu et al.
British journal of pharmacology, 176(11), 1680-1699 (2019-02-15)
An up-regulation of COX-2 in malignant gliomas causes excessive synthesis of PGE2 , which is thought to facilitate brain tumour growth and invasion. However, which downstream PGE2 receptor subtype (i.e., EP1 -EP4 ) directly contributes to COX activity-promoted glioma growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service