Skip to Content
Merck
All Photos(1)

Key Documents

SML1206

Sigma-Aldrich

RA-9

≥98% (HPLC)

Synonym(s):

(3E,5E)-3,5-Bis[(4-Nitrophenyl)methylene]-4-piperidinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€95.20
25 MG
€372.00

€95.20


Check Cart for Availability


Select a Size

Change View
5 MG
€95.20
25 MG
€372.00

About This Item

Empirical Formula (Hill Notation):
C19H15N3O5
CAS Number:
Molecular Weight:
365.34
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€95.20


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(/C(CNC/1)=C/C2=CC=C([N+]([O-])=O)C=C2)C1=C\C3=CC=C([N+]([O-])=O)C=C3

InChI

1S/C19H15N3O5/c23-19-15(9-13-1-5-17(6-2-13)21(24)25)11-20-12-16(19)10-14-3-7-18(8-4-14)22(26)27/h1-10,20H,11-12H2/b15-9+,16-10+

InChI key

YUYPWAMLWZVHAE-KAVGSWPWSA-N

Biochem/physiol Actions

RA-9 is a cell permeable and potent inhibitor of the ubiquitin-proteasome system (UPS) that targets deubiquitinating enzymes (DUB) without affecting 20S proteasome catalytic-core activity. RA-9 directly inhibits activity of following DUB UCH-L1, UCH-L3, USP2, USP5 and USP8 deubiquitinating enzymes, which results in rapid accumulation of poly-ubiquitinated proteins leading to cell cycle checkpoint arrest and tumor cell apoptosis.
RA-9 is a cell permeable and potent inhibitor of the ubiquitin-proteasome system (UPS) that targets deubiquitinating enzymes (DUB).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service