Skip to Content
Merck
All Photos(1)

Key Documents

SML1189

Sigma-Aldrich

4-P-PDOT

≥98% (HPLC)

Synonym(s):

4-Phenyl-2-propionamidotetralin, 4P-PDOT, AH 024, N-(1,2,3,4-Tetrahydro-4-phenyl-2-naphthalenyl)-propanamide, cis-4-Phenyl-2-propionamidotetralin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€97.60
25 MG
€296.00

€97.60


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€97.60
25 MG
€296.00

About This Item

Empirical Formula (Hill Notation):
C19H21NO
CAS Number:
Molecular Weight:
279.38
UNSPSC Code:
12352200
NACRES:
NA.77

€97.60


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

N(C1CC(c3c(cccc3)C1)c2ccccc2)C(=O)CC

InChI

1S/C19H21NO/c1-2-19(21)20-16-12-15-10-6-7-11-17(15)18(13-16)14-8-4-3-5-9-14/h3-11,16,18H,2,12-13H2,1H3,(H,20,21)

InChI key

RCYLUNPFECYGDW-UHFFFAOYSA-N

Biochem/physiol Actions

4-P-PDOT is a slelctive MT2 melatonin receptor antagonist 300 fold selective for MT2 (MEL-1B-R) vs MT1 (MEL-1A-R) melatonin receptor subtype.
4-P-PDOT is a slelctive MT2 melatonin receptor antagonist.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujuan Wang et al.
PeerJ, 9, e10627-e10627 (2021-03-19)
Granulosa cells play an essential physiological role in mediating the follicle development and survival or apoptosis of granulosa cells dictate the follicle development or atresia. The aim of this study was to investigate the role of high dose (10-5 M)
Jun Sung Kim et al.
Stem cell research & therapy, 12(1), 114-114 (2021-02-07)
Retarded wound healing in patients with obesity contributes to a risk of complications associated with vascular insufficiency and oxidative stress. The high cholesterol levels of patients with obesity are associated with apoptosis of engrafted umbilical cord blood-derived mesenchymal stem cells
Jing-Li Tao et al.
Antioxidants (Basel, Switzerland), 10(2) (2021-02-03)
In mammalian ovaries, the avascular environment within follicular cavity is supposed to cause hypoxic status in granulosa cells (GCs), leading to apoptotic cell death accompanied by cumulative reactive oxygen species (ROS) production. Melatonin (N-acetyl-5-methoxytryptamine, MT), a broad-spectrum antioxidant that exists
Jingli Tao et al.
Autophagy, 14(11), 1850-1869 (2018-07-10)
To explore the anti-inflammatory activity of endogenous produced melatonin, a melatonin-enriched animal model (goat) with AANAT transfer was successfully generated with somatic cell nuclear transfer (SCNT) technology. Basically, a pIRES2-EGFP-AANAT expression vector was constructed and was transferred into the female

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service