Skip to Content
Merck
All Photos(1)

Key Documents

SML1183

Sigma-Aldrich

BMH-21

≥98% (HPLC)

Synonym(s):

N-[2-(Dimethylamino)ethyl]-12-oxo-2H-benzo[g]pyrido[2,1-b]quinazoline-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€109.00
25 MG
€438.00

€109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€109.00
25 MG
€438.00

About This Item

Empirical Formula (Hill Notation):
C21H20N4O2
CAS Number:
Molecular Weight:
360.41
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€109.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

faint yellow to dark orange

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(N(C=CC=C1C(NCCN(C)C)=O)C1=N2)C(C2=C3)=CC4=C3C=CC=C4

InChI

1S/C21H20N4O2/c1-24(2)11-9-22-20(26)16-8-5-10-25-19(16)23-18-13-15-7-4-3-6-14(15)12-17(18)21(25)27/h3-8,10,12-13H,9,11H2,1-2H3,(H,22,26)

InChI key

BXYDVWIAGDJBEC-UHFFFAOYSA-N

General description

BMH-21 is a planar heterocyclic small molecule DNA intercalator.[1]

Application

BMH-21 has been used in cell culture.[2]

Biochem/physiol Actions

BMH-21 has the ability to bind ribosomal DNA and prevent RNA polymerase I (Pol I) transcription and results in the disintegration of the nucleolus.[1][3] It does not impair DNA damage detection.[1] BMH-21 is considered as an important activator of the p53 pathway.[3]
BMH-21 is a potent inhibitor of RNA Pol I.
BMH-21 is a potent inhibitor of RNA Pol I. BMH-21 binds strongly to GC-rich DNA sequences, ultimately inhibiting RNA Pol I, blocking transcription and disrupting the nucleolar structure. BMH-1 causes dissociation of the RPA194 catalytic subunit from Pol I, and disassembly of Pol I:DNA complexes. The compound BMH-21 inhibits proliferation of a broad range of tumor cell lines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BMH-21 inhibits viability and induces apoptosis by p53-dependent nucleolar stress responses in SKOV3 ovarian cancer cells.
Fu X, et al.
Oncology Reports, 38(2), 859-865 (2017)
Leah Bury et al.
eLife, 9 (2020-11-12)
Although originally thought to be silent chromosomal regions, centromeres are instead actively transcribed. However, the behavior and contributions of centromere-derived RNAs have remained unclear. Here, we used single-molecule fluorescence in-situ hybridization (smFISH) to detect alpha-satellite RNA transcripts in intact human
A targeting modality for destruction of RNA polymerase I that possesses anticancer activity.
Peltonen K, et al.
Cancer Cell, 25(1), 77-90 (2014)
Lisa M Ogawa et al.
Molecular biology of the cell, 32(9), 956-973 (2021-03-11)
Nucleoli are dynamic nuclear condensates in eukaryotic cells that originate through ribosome biogenesis at loci that harbor the ribosomal DNA. These loci are known as nucleolar organizer regions (NORs), and there are 10 in a human diploid genome. While there
Dimitris C Kanellis et al.
Science advances, 7(32) (2021-08-06)
Eukaryotic initiation factor 4A-III (eIF4A3), a core helicase component of the exon junction complex, is essential for splicing, mRNA trafficking, and nonsense-mediated decay processes emerging as targets in cancer therapy. Here, we unravel eIF4A3's tumor-promoting function by demonstrating its role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service